Skip to Content
Merck
All Photos(2)

Key Documents

804185

Sigma-Aldrich

SnAP 2-Spiro-(2-Pyr) M Reagent

Synonym(s):

tert-butyl 3-(aminomethyl)-3-((tributylstannyl)methoxy) pyrrolidine-1-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H48N2O3Sn
Molecular Weight:
519.35
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

description

Fp >230°F

Quality Level

form

powder

mp

235 °C

density

1.143 at 25 °C

storage temp.

−20°C

SMILES string

CCCC[Sn](CCCC)(COC1(CN(C(OC(C)(C)C)=O)CC1)CN)CCCC

InChI

1S/C11H21N2O3.3C4H9.Sn/c1-10(2,3)16-9(14)13-6-5-11(7-12,8-13)15-4;3*1-3-4-2;/h4-8,12H2,1-3H3;3*1,3-4H2,2H3;

InChI key

FITIMZOFKJTPHZ-UHFFFAOYSA-N

Application

SnAP Reagents provide a one-step route, in tandem with various aldehyde substrates, to saturated N-heterocycles. The synthesis of N-heterocycles through SnAP Reagents requires mild reaction conditions, and aldehydes bearing aryl, heteroaryl, glyoxyl, aliphatic, and halogenated groups are well tolerated. This product was introduced in collaboration with the Bode Research Group

Automate your N-heterocycle formation with Synple Automated Synthesis Platform (SYNPLE-SC002)

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service