Skip to Content
Merck
All Photos(2)

Key Documents

804142

Sigma-Aldrich

SnAP 3-Spiro-(4-Pip) M Reagent

Synonym(s):

tert-butyl 4-amino-4-(((tributylstannyl)methoxy)methyl) piperidine-1-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H50N2O3Sn
Molecular Weight:
533.38
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

storage temp.

−20°C

SMILES string

CCCC[Sn](CCCC)(COCC1(CCN(C(OC(C)(C)C)=O)CC1)N)CCCC

InChI

1S/C12H23N2O3.3C4H9.Sn/c1-11(2,3)17-10(15)14-7-5-12(13,6-8-14)9-16-4;3*1-3-4-2;/h4-9,13H2,1-3H3;3*1,3-4H2,2H3;

InChI key

XZUSOLFDFJLEMZ-UHFFFAOYSA-N

General description

SnAP (Sn amino protocol) reagents have been developed by Professor Dr. Jeffrey Bode and co-workers. (These reagents are useful for the preparation of medium-ring (6-9 membered) saturated N-heterocycles, including bicyclic and spirocyclic structures.) SnAP reagents are stable and easy to handle in laboratories. They can be prepared from simple raw materials.

Application

SnAP Reagents provide a one-step route, in tandem with various aldehyde substrates, to saturated N-heterocycles. The synthesis of N-heterocycles through SnAP Reagents requires mild reaction conditions, and aldehydes bearing aryl, heteroaryl, glyoxyl, aliphatic, and halogenated groups are well tolerated. This product was introduced in collaboration with the Bode Research Group
SnAP 3-Spiro-(4-Pip) M Reagent may be used in the preparation of unprotected, saturated N-heterocyclic compounds.

Automate your N-heterocycle formation with Synple Automated Synthesis Platform (SYNPLE-SC002)

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SnAP Reagents for a Cross-Coupling Approach to the One-Step Synthesis of Saturated N-Heterocycles.
Luescher MU, et al.
Aldrichimica Acta, 48(2), 43-48 (2015)
Cam-Van T Vo et al.
Nature chemistry, 6(4), 310-314 (2014-03-22)
Interest in saturated N-heterocycles as scaffolds for the synthesis of bioactive molecules is increasing. Reliable and predictable synthetic methods for the preparation of these compounds, especially medium-sized rings, are limited. We describe the development of SnAP (Sn amino protocol) reagents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service