Skip to Content
Merck
All Photos(3)

Key Documents

746061

Sigma-Aldrich

Difluoromethyl 2-pyridyl sulfone

97% (HPLC)

Synonym(s):

2-[(Difluoromethyl)sulfonyl]pyridine, Hu Reagent, 2-(Difluoromethylsulfonyl)pyridine, 2-PySO2CF2H

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5F2NO2S
CAS Number:
Molecular Weight:
193.17
Beilstein:
20317604
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97% (HPLC)

form

solid

mp

44-49 °C

SMILES string

O=S(C1=CC=CC=N1)(C(F)F)=O

InChI

1S/C6H5F2NO2S/c7-6(8)12(10,11)5-3-1-2-4-9-5/h1-4,6H

InChI key

YRQNSTAWTLXCEZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Difluoromethyl 2-pyridyl sulfone (2-PySO2CF2H) is a reagent used in the gem-difluoroolefination of aldehydes and ketones. It is also used as a reagent in the nucleophilic difluoro(sulfonato)methylation of alcohols, N-sulfinyl imines, and halides.

Application

Reagent is used in the olefination of ketones and aldehydes to form gem-difluoro olefins under basic conditions. Product is also used as a crucial intermediate toward making 1,1-difluorinated alkyl chains for the alkylation of heterocycles.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Difluoromethyl 2-pyridyl sulfone: a versatile carbonyl gem-difluoroolefination reagent
Gao B, et al.
Organic Chemistry Frontiers : An International Journal of Organic Chemistry / Royal Society of Chemistry, 2(2), 163-168 (2015)
Difluoromethyl 2?Pyridyl Sulfone
e-EROS Encyclopedia of Reagents for Organic Synthesis (2016)
Difluoromethyl 2?Pyridyl Sulfone
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2016)
Yanchuan Zhao et al.
Organic letters, 12(7), 1444-1447 (2010-03-10)
Difluoromethyl 2-pyridyl sulfone, a previously unknown compound, was found to act as a novel and efficient gem-difluoroolefination reagent for both aldehydes and ketones. It was found that the fluorinated sulfinate intermediate in the reaction is relatively stable, which can be
From difluoromethyl 2-pyridyl sulfone to difluorinated sulfonates: a protocol for nucleophilic difluoro(sulfonato)methylation.
G K Surya Prakash et al.
Angewandte Chemie (International ed. in English), 50(11), 2559-2563 (2011-03-04)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service