Skip to Content
Merck
All Photos(1)

Documents

730327

Sigma-Aldrich

Poly(ethylene glycol) methyl ether methacrylate

average Mn 4,000, methacrylate, methoxy, ≤300 ppm MEHQ as inhibitor

Synonym(s):

Polyethylene glycol, Methoxy PEG methacrylate, Methoxy poly(ethylene glycol) monomethacrylate, Poly(ethylene glycol) monomethyl ether monomethacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CCH3CO2(CH2CH2O)nCH3
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

product name

Poly(ethylene glycol) methyl ether methacrylate, average Mn 4,000, contains ≤300 ppm monomethyl ether hydroquinone as inhibitor

form

powder or crystals

mol wt

average Mn 4,000

contains

≤300 ppm monomethyl ether hydroquinone as inhibitor

reaction suitability

reagent type: chemical modification reagent
reaction type: Polymerization Reactions

transition temp

Tm 56-61 °C

density

1.100 g/cm3

Mw/Mn

<1.1

Ω-end

methacrylate

α-end

methoxy

polymer architecture

shape: linear
functionality: monofunctional

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Synthesized with an initial concentration of ≤5,000 ppm MEHQ

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service