Skip to Content
Merck
All Photos(1)

Key Documents

724033

Sigma-Aldrich

Ethylamine

99.0%

Synonym(s):

Aminoethane, Monoethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5NH2
CAS Number:
Molecular Weight:
45.08
Beilstein:
505933
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22

vapor density

1.56 (15 °C, vs air)

Quality Level

vapor pressure

874 mmHg ( 20 °C)

Assay

99.0%

form

gas

autoignition temp.

721 °F

expl. lim.

14 %
3.5-14 %

bp

16.6 °C (lit.)

mp

-81 °C (lit.)

density

0.680 g/mL at 20 °C (lit.)
0.689 g/mL at 25 °C (lit.)

SMILES string

CCN

InChI

1S/C2H7N/c1-2-3/h2-3H2,1H3

InChI key

QUSNBJAOOMFDIB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Supplied in a Sure/Pac cylinder and has a 316 stainless steel needle valve with a male 1/4" NPTF outlet thread installed . Before using the cylinder, ensure that the valve is closed, then remove the galvanized steel hex cap that seals the outlet valve.

Compatible with the following:

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

also commonly purchased with this product

Product No.
Description
Pricing

hose barb

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Gas 1 - Press. Gas Liquefied gas - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

-34.6 °F - closed cup

Flash Point(C)

-37 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Paige E Mahaney et al.
Journal of medicinal chemistry, 51(13), 4038-4049 (2008-06-19)
Further exploration of the cycloalkanol ethylamine scaffold, of which venlafaxine ( 1) is a member, was undertaken to develop novel and selective norepinephrine reuptake inhibitors (NRIs) for evaluation in a variety of predictive animal models. These efforts led to the
Stacey F Bent et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(3), 956-960 (2010-11-12)
Surface functionalization of semiconductors has been the backbone of the newest developments in microelectronics, energy conversion, sensing device design, and many other fields of science and technology. Over a decade ago, the notion of viewing the surface itself as a
Riccardo Di Corato et al.
ACS nano, 5(2), 1109-1121 (2011-01-12)
Trifunctional polymer nanobeads are prepared by destabilization of a mixture of magnetic nanoparticles, quantum dots, and an amphiphilic polymer, followed by functionalization of the bead surface with folic acid molecules. The distribution of the nanoparticles within the nanobeads can be
Veronica Sandgren et al.
Bioorganic & medicinal chemistry, 20(14), 4377-4389 (2012-06-16)
A series of P1-P3 linked macrocyclic BACE-1 inhibitors containing a hydroxyethylamine (HEA) isostere scaffold has been synthesized. All inhibitors comprise a toluene or N-phenylmethanesulfonamide P2 moiety. Excellent BACE-1 potencies, both in enzymatic and cell-based assays, were observed in this series
Câline Christine et al.
Organic & biomolecular chemistry, 10(46), 9183-9190 (2012-10-23)
The synthesis of a phosphonated acyclic bifunctional chelate L* for the labeling of biomaterial is described. L* is based on a pyridine backbone, functionalized in ortho positions by aminomethyl-bis-methylphosphonic acids, and, in the para position, by a side chain containing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service