708682
RuCl[(R,R)-TsDPEN](mesitylene)
90%
Synonym(s):
[N-[(1R,2R)-2-(Amino-κN)-1,2- diphenylethyl]-4-methylbenzenesulfonamidato-κN]chloro [(1,2,3,4,5,6-η)-1,3,5-trimethylbenzene]-ruthenium
About This Item
Recommended Products
Assay
90%
form
solid
mp
241-249 °C
storage temp.
2-8°C
SMILES string
Cc1cc(C)cc(C)c1.Cc2ccc(cc2)S(=O)(=O)N([Ru]Cl)[C@@H]([C@H](N)c3ccccc3)c4ccccc4
InChI
1S/C21H21N2O2S.C9H12.ClH.Ru/c1-16-12-14-19(15-13-16)26(24,25)23-21(18-10-6-3-7-11-18)20(22)17-8-4-2-5-9-17;1-7-4-8(2)6-9(3)5-7;;/h2-15,20-21H,22H2,1H3;4-6H,1-3H3;1H;/q-1;;;+2/p-1/t20-,21-;;;/m1.../s1
InChI key
XBNBOGZUDCYNOJ-AGEKDOICSA-M
Application
- (2R,3S)-4-Benzyl-2-(hydroxyphenylmethyl)morpholin-3-one from racemic N-benzyl-2-benzoyl-3-morpholinone via dynamic kinetic resolution-based asymmetric transfer hydrogenation (ATH).
- Chiral nonracemic δ-phenylseleno alcohols from δ-phenylseleno ketones via catalytic ATH reaction.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service