Skip to Content
Merck
All Photos(2)

Key Documents

692255

Sigma-Aldrich

RuCl2[(R)−DM−BINAP][(R)−DAIPEN]

Synonym(s):

Dichloro[(R)−2,2′−bis[di(3,5−xylyl)phosphino]−1,1′−binaphthyl][(2R)−1,1−bis(4−methoxyphenyl)−3−methyl−1,2−butanediamine]ruthenium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C71H74Cl2N2O2P2Ru
CAS Number:
Molecular Weight:
1221.28
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

form

powder

mp

197-211 °C

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)C2([NH2][Ru]3(Cl)(Cl)([NH2][C@@H]2C(C)C)[PH](c4cc(C)cc(C)c4)(c5cc(C)cc(C)c5)c6ccc7ccccc7c6-c8c(ccc9ccccc89)[PH]3(c%10cc(C)cc(C)c%10)c%11cc(C)cc(C)c%11)c%12ccc(OC)cc%12

InChI

1S/C52H48P2.C19H26N2O2.2ClH.Ru/c1-33-21-34(2)26-43(25-33)53(44-27-35(3)22-36(4)28-44)49-19-17-41-13-9-11-15-47(41)51(49)52-48-16-12-10-14-42(48)18-20-50(52)54(45-29-37(5)23-38(6)30-45)46-31-39(7)24-40(8)32-46;1-13(2)18(20)19(21,14-5-9-16(22-3)10-6-14)15-7-11-17(23-4)12-8-15;;;/h9-32H,1-8H3;5-13,18H,20-21H2,1-4H3;2*1H;/t;18-;;;/m.1.../s1

InChI key

VMSNEARTLSFOHB-OEGAAENXSA-N

Application

Catalyst for:
  • Noyori-type enantioselective ketone hydrogenation
  • Enantioselective and asymmetric hydrogenation of ketones

Legal Information

Sold in collaboration with Takasago for research purposes only. JP Registration No. 2041996, 2731377, 2935453 JP Application No. 19970359654

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service