Skip to Content
Merck
All Photos(1)

Key Documents

663859

Sigma-Aldrich

Sodium acetate-13C2

endotoxin tested, 99 atom % 13C

Synonym(s):

Acetic acid-13C2 sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH313CO2Na
CAS Number:
Molecular Weight:
84.02
Beilstein:
4586604
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

Quality Level

mp

>300 °C (dec.) (lit.)

suitability

endotoxin tested

mass shift

M+2

SMILES string

[Na+].[13CH3][13C]([O-])=O

InChI

1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1/i1+1,2+1;

InChI key

VMHLLURERBWHNL-AWQJXPNKSA-M

Looking for similar products? Visit Product Comparison Guide

Application

For R&D use only. Not for drug,household, or other uses. Any other use is the sole responsibility of the end user.


Endotoxin testing is done on the bulk form of the product before subdivisionpackaging. This bulk test does not guarantee that the subdivision or repackagedaliquot is endotoxin-free when it is received or used by the customer and doesnot imply suitability for any particular purpose. If the product must beendotoxin-free for the intended application, the product should be tested priorto actual use.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mirko Basen et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(49), 17618-17623 (2014-11-05)
Bioethanol production is achieved by only two metabolic pathways and only at moderate temperatures. Herein a fundamentally different synthetic pathway for bioalcohol production at 70 °C was constructed by insertion of the gene for bacterial alcohol dehydrogenase (AdhA) into the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service