Skip to Content
Merck
All Photos(3)

Documents

663727

Sigma-Aldrich

2,2,2-Trichloroethoxysulfonamide

97%

Synonym(s):

2,2,2-Trichloroethyl sulfamate, Sulfamic acid, 2,2,2-trichloroethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H4Cl3NO3S
CAS Number:
Molecular Weight:
228.48
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

52-58 °C

SMILES string

NS(=O)(=O)OCC(Cl)(Cl)Cl

InChI

1S/C2H4Cl3NO3S/c3-2(4,5)1-9-10(6,7)8/h1H2,(H2,6,7,8)

InChI key

VOKONKGVTXWZJI-UHFFFAOYSA-N

Gene Information

human ... STS(412)

Application

Nitrogen source in an intermolecular rhodium-catalyzed (662623) amination of C-H bonds.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristin Williams Fiori et al.
Journal of the American Chemical Society, 129(3), 562-568 (2007-01-18)
Reaction methodology for intermolecular C-H amination of benzylic and 3 degrees C-H bonds is described. This process uses the starting alkane as the limiting reagent, gives optically pure tetrasubstituted amines through stereospecific insertion into enantiomeric 3 degrees centers, displays high

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service