Skip to Content
Merck
All Photos(1)

Key Documents

606731

Sigma-Aldrich

Iodomethane-12C

≥99.9 atom % 12C, ≥99% (CP), contains copper as stabilizer

Synonym(s):

Methyl-12C iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
12CH3I
CAS Number:
Molecular Weight:
141.93
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.12

description

13C-depleted

isotopic purity

≥99.9 atom % 12C

Assay

≥99% (CP)

contains

copper as stabilizer

refractive index

n20/D 1.529

bp

42 °C (lit.)

mp

-66.5 °C (lit.)

density

2.28 g/mL at 25 °C

application(s)

agriculture
environmental

mass shift

depleted

storage temp.

2-8°C

SMILES string

[12CH3]I

InChI

1S/CH3I/c1-2/h1H3/i1+0

InChI key

INQOMBQAUSQDDS-IGMARMGPSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Riet Bammens et al.
Glycobiology, 25(6), 669-682 (2015-01-23)
The congenital disorders of glycosylation (CDG), a group of inherited diseases characterized by aberrant glycosylation, encompass a wide range of defects, including glycosyltransferases, glycosidases, nucleotide-sugar transporters as well as proteins involved in maintaining Golgi architecture, pH and vesicular trafficking. Mutations
Joel E Schmidt et al.
Angewandte Chemie (International ed. in English), 53(32), 8372-8374 (2014-06-26)
Crystalline molecular sieves are used in numerous applications, where the properties exploited for each technology are the direct consequence of structural features. New materials are typically discovered by trial and error, and in many cases, organic structure-directing agents (OSDAs) are
Hagay Kohay et al.
Environmental science & technology, 49(15), 9280-9288 (2015-07-01)
The presence of nanoconcentrations of persistent pharmaceuticals in treated wastewater effluent and in surface water has been frequently reported. A novel organic-inorganic hybrid sorbent based on adsorbing quarternized poly vinylpyridinium-co-styrene (QPVPcS) to montmorillonite (MMT) was designed for the removal of
Ann-Charlotte Almstrand et al.
Analytical and bioanalytical chemistry, 407(17), 5045-5052 (2015-03-05)
Lipidomic analysis of the complex mixture of lipids isolated from biological systems can be a challenging process that often involves tandem mass spectrometry and interpretation of both precursor ions and product ions relative to the molecular structure of the lipids.
Xiangfeng Meng et al.
Applied microbiology and biotechnology, 99(14), 5885-5894 (2015-01-15)
Glucansucrases are exclusively found in lactic acid bacteria and synthesize a variety of α-glucans from sucrose. They are large multidomain enzymes belonging to the CAZy family 70 of glycoside hydrolase enzymes (GH70). The crystal structure of the N-terminal truncated GTF180

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service