Skip to Content
Merck
All Photos(1)

Key Documents

606340

Sigma-Aldrich

Pentachlorophenol-13C6

99 atom % 13C, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13C6Cl5OH
Molecular Weight:
272.29
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

Quality Level

Assay

98% (CP)

application(s)

agriculture

mass shift

M+6

SMILES string

O[13c]1[13c](Cl)[13c](Cl)[13c](Cl)[13c](Cl)[13c]1Cl

InChI

1S/C6HCl5O/c7-1-2(8)4(10)6(12)5(11)3(1)9/h12H/i1+1,2+1,3+1,4+1,5+1,6+1

InChI key

IZUPBVBPLAPZRR-IDEBNGHGSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hanh Thi Duong et al.
Chemosphere, 219, 784-795 (2018-12-21)
Vietnam's rapid economic development has resulted in dramatic increases in construction and the number of transportation vehicles. There is now growing public concern regarding increasing air pollution, especially in big cities; however, little information is available on air quality, particularly
M Homolková et al.
Environmental science and pollution research international, 23(2), 1408-1413 (2015-09-16)
The production and use of chlorophenolic compounds in industry has led to the introduction of many xenobiotics, among them chlorophenols (CPs), into the environment. Five CPs are listed in the priority pollutant list of the U.S. EPA, with pentachlorophenol (PCP)
Luca Amendola et al.
Analytica chimica acta, 975, 70-77 (2017-05-30)
Tributyltin (TBT) and pentachlorophenol (PCP) are listed as priority substances that warrant monitoring in the European Water Framework Directive (WFD 2000/60/CE), since TBT and PCP were identified as environmental endocrine disruptors and exhibit considerable toxicity toward both aquatic organisms and
Lukas Schmidt et al.
Analytica chimica acta, 965, 123-130 (2017-04-04)
The determination of chlorophenols in urine is an established approach for the assessment of human exposure to these ubiquitous environmental pollutants. However, an analytical procedure which enables the separate determination of all components of this substance group was still lacking.
Adrià Rubirola et al.
Journal of chromatography. A, 1493, 64-75 (2017-03-21)
This paper reports the development of a fully multiresidue and automated on-line solid phase extraction (SPE) - liquid chromatography tandem mass spectrometry (LC-MS/MS) method for the determination of 24 priority substances (PS) belonging to different classes (pesticides, hormones or pharmaceuticals)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service