Skip to Content
Merck
All Photos(1)

Key Documents

602183

Sigma-Aldrich

Xenon-132Xe

60 atom %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
132Xe
CAS Number:
Molecular Weight:
131.90
MDL number:
UNSPSC Code:
12142001
PubChem Substance ID:

isotopic purity

60 atom %

Assay

≥99.5% (Total Xenon)

form

gas

impurities

<0.5% Total others

bp

-107.1 °C (lit.)

mass shift

depleted

SMILES string

[132Xe]

InChI

1S/Xe/i1+1

InChI key

FHNFHKCVQCLJFQ-OUBTZVSYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arghya Chakraborty et al.
The Journal of chemical physics, 143(8), 084312-084312 (2015-09-04)
The electronic transitions of 9-fluorenone FL(+) and 2,3,6,7-dibenzotropone DBT(+) cations were detected in 6 K neon matrices following a mass-selective deposition. The absorptions at 649.2 and 472.2 nm are assigned to the 2 (2)B1←X̃(2)A2 FL(+) and 2(2)A(')←X̃(2)A(') DBT(+) transitions. Absorption spectra of
Mu He et al.
Magnetic resonance imaging, 33(7), 877-885 (2015-05-06)
The aim of this study was to evaluate the effect of hyperpolarized (129)Xe dose on image signal-to-noise ratio (SNR) and ventilation defect conspicuity on both multi-slice gradient echo and isotropic 3D-radially acquired ventilation MRI. Ten non-smoking older subjects (ages 60.8±7.9years)
Eladio Rodriguez-Diaz et al.
Inflammatory bowel diseases, 20(6), 1029-1036 (2014-05-07)
In 10% to 15% of individuals, inflammatory bowel disease (IBD) is difficult to classify as ulcerative colitis (UC) or Crohn's disease (CD). Previous work has demonstrated that probe-based elastic scattering spectroscopy (ESS) can produce spectra, informed by parameters like tissue
Mathias Steglich et al.
The Journal of chemical physics, 142(24), 244311-244311 (2015-07-03)
The 1(3)Σu (-)←X(3)Σg (-) transition of linear HC5H (A) has been observed in a neon matrix and gas phase. The assignment is based on mass-selective experiments, extrapolation of previous results of the longer HC2n+1H homologues, and density functional and multi-state
Jacques H Abraini et al.
Anesthesiology, 121(5), 1018-1027 (2014-09-12)
The mechanisms by which general anesthetics, including xenon and nitrous oxide, act are only beginning to be discovered. However, structural approaches revealed weak but specific protein-gas interactions. To improve knowledge, we performed x-ray crystallography studies under xenon and nitrous oxide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service