Skip to Content
Merck
All Photos(1)

Documents

601985

Sigma-Aldrich

Krypton-84Kr

90 atom %

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
84Kr
CAS Number:
Molecular Weight:
83.91
MDL number:
UNSPSC Code:
12142003
PubChem Substance ID:

isotopic purity

90 atom %

Assay

≥99.5% (Total Krypton)

form

gas

impurities

<0.5% Total others

mass shift

depleted

SMILES string

[84Kr]

InChI

1S/Kr/i1+0

InChI key

DNNSSWSSYDEUBZ-IGMARMGPSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Compr. Gas

Storage Class Code

2B - Aerosol cans and lighters

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xavier Devaux et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 16(12), 2692-2701 (2015-07-03)
The results of a study on the evolution of HiPco single-walled carbon nanotubes during the oxidizing action of H2 SO4 and HNO3 are presented. The process conditions used have been chosen so as to avoid any significant damage to the
Gamil A Guirgis et al.
The journal of physical chemistry. A, 119(47), 11532-11547 (2015-10-06)
The FT-microwave spectrum (6.5-26 GHz) of (chloromethyl)fluorosilane (ClCH2-SiH2F) has been recorded and 250 transitions for the parent species along with (13)C, (37)Cl, (29)Si, and (30)Si isotopologues have been assigned for trans conformer. Infrared spectra (3100 to 400 cm(-1)) of gas
James G Carter et al.
Angiogenesis, 18(1), 23-30 (2014-10-03)
Anti-VEGF-A therapy has become a mainstay of treatment for ocular neovascularisation and in cancer; however, their effectiveness is not universal, in some cases only benefiting a minority of patients. Anti-VEGF-A therapies bind and block both pro-angiogenic VEGF-Axxx and the partial
Charlotte Michaux et al.
Infection and immunity, 82(9), 3599-3611 (2014-06-11)
Candidate small RNAs (sRNAs) have recently been identified in Enterococcus faecalis, a Gram-positive opportunistic pathogen, and six of these candidate sRNAs with unknown functions were selected for a functional study. Deletion mutants and complemented strains were constructed, and their virulence
Yvonne Kohlmann et al.
Journal of proteome research, 13(10), 4325-4338 (2014-09-10)
Ralstonia eutropha H16 is a denitrifying microorganism able to use nitrate and nitrite as terminal electron acceptors under oxygen deprivation. To identify proteins showing an altered expression pattern in response to oxygen supply, R. eutropha cells grown aerobically and anaerobically

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service