Skip to Content
Merck
All Photos(1)

Key Documents

550809

Sigma-Aldrich

Phenylguanidine carbonate salt

≥99%

Synonym(s):

1-Phenylguanidine carbonate, N-Phenylguanidine carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHC(=NH)NH2·(H2CO3)x
CAS Number:
Molecular Weight:
135.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

mp

149-153 °C (lit.)

SMILES string

OC(O)=O.NC(=N)Nc1ccccc1

InChI

1S/C7H9N3.CH2O3/c8-7(9)10-6-4-2-1-3-5-6;2-1(3)4/h1-5H,(H4,8,9,10);(H2,2,3,4)

InChI key

XDSYAIICRRZSJX-UHFFFAOYSA-N

Application

Phenylguanidine carbonate salt may be used in the preparation of the following bioactive compounds:
  • (E)-methyl 2-(2-((6-cyclopropyl-2-(phenylamino)pyrimidin-4-yloxy)methyl)phenyl)-3-methoxyacrylate
  • cyprodinil
  • 5,6-dihydropyrido[2,3-d]pyrimidine
  • ethyl 1-methyl-8-(phenylamino)-4,5-dihydro-1H-pyrazolo[4,3-h]quinazoline-3-carboxylate
  • 2-(phenylamino)-4-(trifluoromethyl)-1,6-dihydropyrimidine derivatives
Reactant involved in synthesis of polo-like kinase 1 inhibitors

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and SAR of new pyrazolo [4, 3-h] quinazoline-3-carboxamide derivatives as potent and selective MPS1 kinase inhibitors.
Caldarelli M, et al.
Bioorganic & Medicinal Chemistry Letters, 21(15), 4507-4511 (2011)
Synthesis of bactericide Cyprodinil.
Qu J, et al.
Zhejiang Gongye Daxue Xuebao (Journal of Zhejiang University of Technology), 1, 7-7 (2012)
Synthesis of (E)-Methyl 2-(2-((6-cyclopropyl-2-(phenylamino) pyrimidin-4-yloxy) methyl) phenyl)-3-methoxyacrylate and Its Fungicidal Activity.
Li HC, et al.
Agrochemicals, 4, 6-6 (2009)
An unusual Michael addition of 3, 3-dimethoxypropanenitrile to 2-aryl acrylates: a convenient route to 4-unsubstituted 5, 6-dihydropyrido [2, 3-d] pyrimidines.
Berzosa X, et al.
The Journal of Organic Chemistry, 75(2), 487-490 (2009)
Solvent-Free Synthesis of 2-(Phenylamino)-4-(trifluoromethyl)-1, 6-dihydropyrimidine Derivatives Catalyzed by Sulfamic Acid.
Shen S, et al.
Journal of Heterocyclic Chemistry, 49(5), 1033-1037 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service