534641
Amano Lipase PS, from Burkholderia cepacia
≥23,000 U/g, pH 7.0, 50 °C (Optimum pH and temperature)
Synonym(s):
APS-BCL, Pseudomonas cepacia (APS-PCL)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
specific activity
≥23,000 U/g, pH 7.0, 50 °C (Optimum pH and temperature)
Application
Amano Lipase PS is generally used in the enantioselective transesterification and hydrolysis. Applications include:
- Lipase catalyzed transesterification of prochiral pyrimidine acyclonucleoside.
- Lipase catalyzed hydrolysis of diacetylated pyrimidine acyclonucleosides.
- Enantiomer selective acylation of racemic alcohols in continuous-flow bioreactors.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Resp. Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Immobilization of lipase from Burkholderia cepacia into calcium carbonate microcapsule and its use for enzymatic reactions in organic and aqueous media.
Journal of Molecular Catalysis. B, Enzymatic, 109, 94-100 (2014)
Lipase-catalyzed enantioselective transesterification of prochiral 1-((1, 3-dihydroxypropan-2-yloxy) methyl)-5, 6, 7, 8-tetrahydroquinazoline-2, 4 (1H, 3H)-dione in ionic liquids.
Chirality, 30(2), 206-214 (2018)
Reverse Stereoselectivity in the Lipase-Catalyzed Hydrolysis of Diacetylated Pyrimidine Acyclonucleosides.
ChemCatChem, 8(23), 3644-3649 (2016)
Enantiomer selective acylation of racemic alcohols by lipases in continuous-flow bioreactors.
Tetrahedron Asymmetry, 19(2), 237-246 (2008)
Chemistry (Weinheim an der Bergstrasse, Germany), 25(43), 10156-10164 (2019-05-29)
A new protocol based on lipase-catalyzed tandem reaction toward α,β-enones/enoesters is presented. For the synthesis of the desired products the tandem process based on enzyme-catalyzed hydrolysis and Knoevenagel reaction starting from enol acetates and aldehyde is developed. The relevant impact
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service