Skip to Content
Merck
All Photos(1)

Key Documents

531197

Sigma-Aldrich

Ethyl 6-chloropyridine-3-carboxylate

97%

Synonym(s):

Ethyl 6-chloronicotinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8ClNO2
CAS Number:
Molecular Weight:
185.61
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

26-30 °C (lit.)

SMILES string

CCOC(=O)c1ccc(Cl)nc1

InChI

1S/C8H8ClNO2/c1-2-12-8(11)6-3-4-7(9)10-5-6/h3-5H,2H2,1H3

InChI key

ILDJJTQWIZLGPO-UHFFFAOYSA-N

General description

Ethyl 6-chloropyridine-3-carboxylate (Ethyl-6-chloronicotinate, E-6-ClN) undergoes direct amidation on reacting with benzylamine in the presence lanthanum trifluoromethanesulfonate La(OTf)3. The structural and physicochemical properties of E-6-ClN have been investigated based on its spectroscopic data, time-dependent density functional theory and density of state diagrams.

Application

Ethyl 6-chloropyridine-3-carboxylate (Ethyl 6-chloronicotinate) may be used in the preparation of ethyl 5-{[5-(1H-benzimidazol-2-yl)pyridin-2-yl]ethynyl}pyridine-2-carboxylate by reacting with 2-[6-(ethynyl)pyridin-3-yl]-1H-benzimidazole under microwave irradiation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Microwave-assisted Sonogashira coupling of novel 2-[6-(arylethynyl) pyridin-3-yl]-1H-benzimidazole derivatives.
Raut CN, et al.
ARKIVOC (Gainesville, FL, United States), 11, 105-114 (2009)
The spectroscopic (FT-IR, FT-Raman, dispersive Raman and NMR) study of ethyl-6-chloronicotinate molecule by combined density functional theory.
Karabacak M, et al.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 153, 754-770 (2016)
Hiroyuki Morimoto et al.
Organic letters, 16(7), 2018-2021 (2014-03-26)
Lanthanum trifluoromethanesulfonate is an effective single-component catalyst for synthesizing a variety of amides directly from esters and amines under mild conditions. Highly selective amidation of esters and amines, as well as catalyst-controlled amidation of esters, demonstrated the effectiveness of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service