Skip to Content
Merck
All Photos(1)

Key Documents

525286

Sigma-Aldrich

Sucrose acetate isobutyrate solution

90 wt. % in denatured ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H62O19
CAS Number:
Molecular Weight:
846.91
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

40 mmHg ( 20 °C)

Quality Level

concentration

90 wt. % in denatured ethanol

bp

105 °C

density

1.1 g/mL at 25 °C

SMILES string

CC(C)C(=O)OC[C@]1(O[C@H](COC(C)=O)[C@@H](OC(=O)C(C)C)[C@@H]1OC(=O)C(C)C)O[C@H]2O[C@H](COC(C)=O)[C@@H](OC(=O)C(C)C)[C@H](OC(=O)C(C)C)[C@H]2OC(=O)C(C)C

InChI

1S/C40H62O19/c1-18(2)33(43)51-17-40(32(57-38(48)23(11)12)29(54-35(45)20(5)6)27(58-40)16-50-25(14)42)59-39-31(56-37(47)22(9)10)30(55-36(46)21(7)8)28(53-34(44)19(3)4)26(52-39)15-49-24(13)41/h18-23,26-32,39H,15-17H2,1-14H3/t26-,27-,28-,29-,30+,31-,32+,39-,40+/m1/s1

InChI key

ZNEBZIJCDDCNRC-SWTLDUCYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

60.6 °F - closed cup

Flash Point(C)

15.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P E Noker et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 24(12), 1287-1293 (1986-12-01)
The disposition of 200 mg/kg of 14C-labelled sucrose octa-isobutyrate (14C-SOIB), a component of sucrose acetate isobutyrate (SAIB), a beverage emulsion stabilizer, was studied in rats, dogs and monkeys. After oral administration of 14C-SOIB to three rats, 3-15% of the dose
A Knoll et al.
Microvascular research, 58(1), 1-9 (1999-07-02)
The diuretic amiloride has been reported to inhibit both Na+-H+ antiport and the urokinase-type plasminogen activator. As a consequence of these inhibitions, neovascularization may also be inhibited. We hypothesized that if amiloride could be effectively delivered in a site-specific manner
J E Nally et al.
Vaccine, 19(4-5), 492-497 (2000-10-12)
Streptococcus equi causes equine strangles, a highly contagious disease of the upper respiratory tract. The antiphagocytic surface protein SeM is strongly immunogenic and evokes mucosal and systemic antibodies during convalescence. The present study investigated the potential of sucrose acetate isobutyrate
R C Reynolds et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 36(2), 81-93 (1998-03-31)
Sucrose acetate isobutyrate (SAIB), a mixture of esters of sucrose with a composition approximating the name sucrose diacetate hexaisobutyrate, has been used for over 30 yr in many countries as a 'weighting' or 'density-adjusting' agent in non-alcoholic carbonated and non-carbonated
Eun Seong Lee et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 29(5), 435-441 (2006-09-19)
This study investigated the suitability of microsphere formulations for extended protein delivery and complete protein release. These microspheres were prepared by a multi-emulsion method and prepared using a mixture of poly(lactide-co-glycolide) (PLGA), RG 502H (lactide:glycolide=50:50, M(W) 9300) and sucrose acetate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service