Skip to Content
Merck
All Photos(1)

Documents

50780

Sigma-Aldrich

Gold(III) chloride hydrate

~52% Au basis

Synonym(s):

Auric chloride, Chloroauric acid, Hydrogen tetrachloroaurate(III) hydrate, Tetrachloroauric(III) acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HAuCl4 · aq
CAS Number:
Molecular Weight:
339.79 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

form

powder

concentration

~52% Au

color

brown

density

3.9 g/mL at 25 °C

SMILES string

O.Cl.Cl[Au](Cl)Cl

InChI

1S/Au.4ClH.H2O/h;4*1H;1H2/q+3;;;;;/p-3

InChI key

OVJUDSKPNIBLOO-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Gold(III) chloride hydrate is a yellowish crystalline solid and a powerful oxidizing agent widely used in gold electroplating, catalyst synthesis, and organic transformations.

Application

Gold(III) chloride hydrate can be used:
  • As a precursor to synthesize blue-emitting gold nanoclusters.
  • As a dopant to enhance the optoelectronic properties of single-walled carbon nanotubes.
  • To produce composite colloids of gold and polypyrrole.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Blue emitting gold nanoclusters templated by poly-cytosine DNA at low pH and poly-adenine DNA at neutral pH
AC Thomas, et al.
Chemical Communications (Cambridge, England), 48, 6845-6847 (2012)
Reaction of pyrrole and chlorauric acid a new route to composite colloids
Matthew C Henry, et al.
Journal of the Electrochemical Society, 148, D155-D155 (2001)
Highly efficient doping of carbon nanotube films with chloroauric acid by dip-coating
Orysia T. Zaremba, et al.
Materials Science and Engineering, B, 278, 115648-115648 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service