Skip to Content
Merck
All Photos(1)

Key Documents

486469

Sigma-Aldrich

Carbonyl-13C sulfide

<5 atom % 18O, 99 atom % 13C

Synonym(s):

Carbon-13C oxysulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13COS
CAS Number:
Molecular Weight:
61.07
MDL number:
UNSPSC Code:
12352119
PubChem Substance ID:
NACRES:
NA.12

vapor density

2.1 (20 °C, vs air)

vapor pressure

9034 mmHg ( 21 °C)

isotopic purity

99 atom % 13C
<5 atom % 18O

bp

−50 °C (lit.)

mp

−138 °C (lit.)

mass shift

M+1

SMILES string

O=[13C]=S

InChI

1S/COS/c2-1-3/i1+1

InChI key

JJWKPURADFRFRB-OUBTZVSYSA-N

Related Categories

Packaging

250 mL quantity is packaged in a 25 mL Sure/Pac cylinder with stainless steel 1/4" male NPT valve.
All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Recommended products

Stainless steel hose adapter Z146838 or stainless steel body mini gas regulator Z513547 is recommended.
PTFE sealing washers GCA inlet size 180 Z146994
PTFE sealing washers CGA inlet size 110 Z146986

Legal Information

Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

hose barb

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Pictograms

Gas cylinderSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Irrit. 2 - Press. Gas Liquefied gas - Skin Irrit. 2 - STOT SE 3

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service