Skip to Content
Merck
All Photos(1)

Documents

47560

Sigma-Aldrich

Fmoc-Orn(Boc)-OH

≥96.0% (HPLC)

Synonym(s):

Nα-Fmoc-Nδ-Boc-L-ornithine, Nδ-Boc-Nα-Fmoc-L-ornithine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H30N2O6
CAS Number:
Molecular Weight:
454.52
Beilstein:
4772025
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Assay

≥96.0% (HPLC)

form

solid

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Boc
Fmoc

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)NCCC[C@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C25H30N2O6/c1-25(2,3)33-23(30)26-14-8-13-21(22(28)29)27-24(31)32-15-20-18-11-6-4-9-16(18)17-10-5-7-12-19(17)20/h4-7,9-12,20-21H,8,13-15H2,1-3H3,(H,26,30)(H,27,31)(H,28,29)/t21-/m0/s1

InChI key

JOOIZTMAHNLNHE-NRFANRHFSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kuo-Yuan Hung et al.
European journal of medicinal chemistry, 136, 154-164 (2017-05-12)
The 20-residue linear peptide A20FMDV2 has been shown to exhibit high selectivity and affinity for the tumour-related αvβ6 integrin and has potential as a vector for therapeutic drugs. However, it exhibits poor half-life in plasma in part due to its
Santanu Mondal et al.
ACS chemical biology, 13(4), 1057-1065 (2018-03-09)
Protein arginine deiminases (PADs) play an important role in the pathogenesis of various diseases, including rheumatoid arthritis, multiple sclerosis, lupus, ulcerative colitis, and breast cancer. Therefore, the development of PAD inhibitors has drawn significant research interest in recent years. Herein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service