Skip to Content
Merck
All Photos(1)

Documents

44570

Sigma-Aldrich

Dowtherm® A

eutectic mixture of 26.5% diphenyl + 73.5% diphenyl oxide

Synonym(s):

Diphyl

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

refractive index

n20/D 1.590

mp

12-14 °C (lit.)

SMILES string

c1ccc(cc1)-c2ccccc2.O(c3ccccc3)c4ccccc4

InChI

1S/C12H10O.C12H10/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;1-3-7-11(8-4-1)12-9-5-2-6-10-12/h1-10H;1-10H

InChI key

MHCVCKDNQYMGEX-UHFFFAOYSA-N

General description

Dowtherm® A is a thermally stable organic heat transfer fluid composed of a eutectic mixture of highly stable compounds, biphenyl, and diphenyl oxide. Dowtherm® A is suitable for high-temperature heat transfer applications up to 400 °C. This fluid can be used in both liquid and vapor phase systems.

Application

Dowtherm® A can be used as a high boiling solvent in cyclization reactions. For example, it is used to synthesize:      
  • 1-Hydroxy-5-methoxy-1,2-dihydrobenzo[f][2,7]naphthyridines from 2-chloro-3-formyl quinolines.     
  • Isoindolinone derivatives via intramolecular Diels–Alder cycloaddition reactions of substituted pyridazines.
  • Phosphorylated azaheterocycles from 2-diethoxyphosphoryl-3-methoxyacrylate via intramolecular cyclizations under thermal conditions.

Legal Information

DOWTHERM is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient synthesis of phosphorylated ortho-fused azaheterocycles
Modranka J and Janecki T
Tetrahedron, 67(49), 9595-9601 (2011)
V M Sheĭbak et al.
Gigiena i sanitariia, (4)(4), 81-82 (2008-12-23)
Experiments on rats were carried out to study the effect of dinil on the animals on chronic intake of the agent in the quantities exceeding the maximum permissible concentrations by many times. Despite few biochemical changes, there was a certain
[2 cases of acute dinil poisoning].
A N Kapustina et al.
Gigiena truda i professional'nye zabolevaniia, (3)(3), 50-51 (1983-03-01)
F O Dorgelo et al.
International archives of occupational and environmental health, 56(2), 129-134 (1985-01-01)
A study was carried out in a nylon producing plant to determine exposure to Dowtherm A from frequently occurring leaks in the heating system. The daily exposure to both diphenyl and diphenylether of 20 workers was determined over seven consecutive
Thermal neutron cross sections and diffusion parameters of diphenyl and Dowtherm-A using a synthetic scattering function.
Florido PC, et al.
Journal of Nuclear Materials, 185(1), 114-122 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service