Skip to Content
Merck
All Photos(1)

Key Documents

445541

Sigma-Aldrich

Hafnium(IV) tert-butoxide

99.99% trace metals basis (purity excludes ~2000 ppm zirconium.)

Synonym(s):

Hafnium tetrakis(tert -butoxide), Hafnium(IV) tert -butoxide, Tetra-tert -butoxyhafnium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Hf[OC(CH3)3]4
CAS Number:
Molecular Weight:
470.94
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99% trace metals basis (purity excludes ~2000 ppm zirconium.)

form

liquid

reaction suitability

core: hafnium

refractive index

n20/D 1.424 (lit.)

bp

90 °C/5 mmHg (lit.)

density

1.166 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)O[Hf](OC(C)(C)C)(OC(C)(C)C)OC(C)(C)C

InChI

1S/4C4H9O.Hf/c4*1-4(2,3)5;/h4*1-3H3;/q4*-1;+4

InChI key

WZVIPWQGBBCHJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Atomic number of base material: 72 Hafnium

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sean W Depner et al.
ACS nano, 8(5), 4678-4688 (2014-04-23)
We demonstrate that the degree of branching of the alkyl (R) chain in a Hf(OR)4 precursor allows for control over the length of HfO2 nanocrystals grown by homocondensation of the metal alkoxide with a metal halide. An extended nonhydrolytic sol-gel

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service