Skip to Content
Merck
All Photos(1)

Key Documents

436038

Sigma-Aldrich

Iron(III) phosphate tetrahydrate

Synonym(s):

Ferric phosphate tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FePO4 · 4H2O
CAS Number:
Molecular Weight:
222.88
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

97-103% (with sodium thiosulphate, titration)

form

powder

composition

Fe, 24%

application(s)

battery manufacturing

SMILES string

O.O.O.O.[Fe+3].[O-]P([O-])([O-])=O

InChI

1S/Fe.H3O4P.4H2O/c;1-5(2,3)4;;;;/h;(H3,1,2,3,4);4*1H2/q+3;;;;;/p-3

InChI key

RTOYJIFECJCKIK-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service