Skip to Content
Merck
All Photos(2)

Key Documents

416428

Sigma-Aldrich

2,2′-Methylenebis[(4S)-4-phenyl-2-oxazoline]

97%

Synonym(s):

(S,S)-2,2′-Methylenebis(4-phenyl-2-oxazoline)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18N2O2
CAS Number:
Molecular Weight:
306.36
Beilstein:
4202633
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

viscous liquid

optical activity

[α]20/D ~−90°, c = 1 in ethanol

refractive index

n20/D 1.586 (lit.)

bp

131-134 °C/0.01 mmHg (lit.)

density

1.28 g/mL at 25 °C (lit.)

SMILES string

C1OC(CC2=N[C@H](CO2)c3ccccc3)=N[C@H]1c4ccccc4

InChI

1S/C19H18N2O2/c1-3-7-14(8-4-1)16-12-22-18(20-16)11-19-21-17(13-23-19)15-9-5-2-6-10-15/h1-10,16-17H,11-13H2/t16-,17-/m1/s1

InChI key

IUFHJPXOLHSJTC-IAGOWNOFSA-N

Application

C2 symmetric ligand for enantioselective catalysis. Easily forms bidentate coordination complexes due to the strong affinity of the oxazoline nitrogen for various metals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gant, T.G. Meyers, A.I.
Tetrahedron, 50, 2297-2297 (1994)
Bolm, C.
Angewandte Chemie (International Edition in English), 30, 542-542 (1991)
Pfaltz, A.
Accounts of Chemical Research, 26, 339-339 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service