416282
Tris-(8-hydroxyquinoline)aluminum
98% trace metals basis
Synonym(s):
8-Hydroxyquinoline aluminum salt, Alq3, Aluminum 8-hydroxyquinolinate, Aluminum oxinate, Tris-(8-hydroxyquinolinato)aluminum
About This Item
Recommended Products
Assay
98% trace metals basis
form
powder
mp
>300 °C (lit.)
Orbital energy
HOMO 5.8 eV
LUMO 3.1 eV
OLED Device Performance
ITO/MoO3/NPD/Alq3/BPhen/LiF/Al
ITO/NPD/CBP:Ir(ppy)3/BCP/Alq3/Mg:Al
ITO/NPD/TCTA/BCPO:Ir(piq)3 (7-8%)/BCP/Alq3/LiF/Al
ITO/NPD/TCTA/BCPO:Ir(ppy)3 (7-8%)/BCP/Alq3/LiF/Al
ITO/m-MTDATA/NPD/TTPhPhB/Alq3/LiF/Al
SMILES string
O([Al](Oc1cccc2cccnc12)Oc3cccc4cccnc34)c5cccc6cccnc56
InChI
1S/3C9H7NO.Al/c3*11-8-5-1-3-7-4-2-6-10-9(7)8;/h3*1-6,11H;/q;;;+3/p-3
InChI key
TVIVIEFSHFOWTE-UHFFFAOYSA-K
Looking for similar products? Visit Product Comparison Guide
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service