Skip to Content
Merck
All Photos(1)

Key Documents

409537

Sigma-Aldrich

Poly(ethylene glycol) methacrylate

average Mn 360, methacrylate, 500-800 ppm MEHQ as inhibitor

Synonym(s):

Ethoxylated 2-hydroxyethyl methacrylate, PEGMA, Poly(ethylene oxide) monomethacrylate, Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)CO(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

Product Name

Poly(ethylene glycol) methacrylate, average Mn 360, contains 500-800 ppm MEHQ as inhibitor

form

liquid

Quality Level

mol wt

average Mn 360

contains

500-800 ppm MEHQ as inhibitor

reaction suitability

reagent type: cross-linking reagent
reaction type: Polymerization Reactions

refractive index

n20/D 1.464

density

1.105 g/mL at 25 °C

Ω-end

hydroxyl

α-end

methacrylate

polymer architecture

shape: linear
functionality: heterobifunctional

SMILES string

O(CCO)C(=O)C(=C)C

InChI

1S/C6H10O3/c1-5(2)6(8)9-4-3-7/h7H,1,3-4H2,2H3

InChI key

WOBHKFSMXKNTIM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Polyethylene glycol (PEG) is a suitable material for biological applications since it does not trigger an immune response. It can by synthesized by anionic ring opening polymerization of ethylene oxide with a range of end groups (such as methacrylate: PEGMA) attached. PEG is a hydrophilic polymer that can form hydrogels when cross linked into networks. Acrylate and methacrylate chain ends undergo chain polymerization to from PEGMA hydrogels.

Application

PEGMA can beused as a monomer to synthesize:
  • Degradable microspheres using asuspension polymerization process. The amphiphilic nature of PEGMA allows performingthe polymerization by direct oil in the water suspension process.
  • Polymeric chelating beads for the effectiveremoval of heavy metals from aqueous solutions.
It can also be used to synthesize micro or nanoparticles-basedchitosan /PEGMA composite by Michael addition reaction. The PEGMA graftingimproves the solubility of chitosan in aqueous media. These particles can beused as carriers in the ophthalmic drug delivery system.

Physical form

Monomethacrylate functionalized

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biofouling-resistance expanded poly (tetrafluoroethylene) membrane with a hydrogel-like layer of surface-immobilized poly (ethylene glycol) methacrylate for human plasma protein repulsions.
Chang Y, et al.
Journal of Membrane Science, 323(1), 77-84 (2008)
Mir Mukkaram Ali;
Macromolecules, 37, 5219-5227 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service