Skip to Content
Merck
All Photos(2)

Key Documents

346152

Sigma-Aldrich

Ethynylmagnesium bromide solution

0.5 M in THF

Synonym(s):

Acetylenemagnesium bromide, Bromoethynylmagnesium, Ethynylmagnesium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC≡CMgBr
CAS Number:
Molecular Weight:
129.24
Beilstein:
3600874
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

reaction suitability

reaction type: Grignard Reaction

concentration

0.5 M in THF

density

0.94 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

Br[Mg]C#C

InChI

1S/C2H.BrH.Mg/c1-2;;/h1H;1H;/q;;+1/p-1

InChI key

HUGJUYPSXULVQQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Ethynylmagnesium bromide solution, also known as bromoethynylmagnesium, is a grignard reagent that is commonly used in the synthesis of polymers, and unsaturated phosphonites.

Application

Ethynylmagnesium bromide solution can be used in:
  • Grafting of aliphatic polymer.
  • Synthesis of anthracene-containing species like 1,8-dichloro-10-(ethynyl)anthracene.
  • The preparation of the alkyne component in an enyne cross-metathesis leading to conjugated dienes.
  • Ethynylation of chiral α-(dibenzylamino) aldehydes.
  • The synthesis of ethynyl aziridines.
  • To prepare alkyl(triethynyl)germanes from tert-alkyltrichlorogermanes and 2-butyltrichlorogermane.
  • The preperation of a key intermediate in the total synthesis of (+)-4-demethoxydaunomycin.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 2

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

-20.2 °F

Flash Point(C)

-29 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Grafting of functionalized polymer on porous silicon surface using Grignard reagent
FZ T,et al.
Applied Surface Science, 421, 82-88 (2017)
An enantioselective formal synthesis of 4-demethoxydaunomycin using the catalytic asymmetric ring opening reaction of meso-epoxide with p-anisidine
Sekine A, et al.
Tetrahedron, 58(1), 75-82 (2002)
Improved Access to 1, 8-Dichloro-10-(ethynyl) anthracene: A Useful Building Block for (Semi-) rigid Organic Frameworks
Jan-Hendrik L,et al.
Synthesis, 50, 2009-2018 (2018)
Tandem enyne metathesis and Claisen rearrangement: a versatile approach to conjugated dienes of variable substitution patterns
Clark DA, et al.
Journal of the American Chemical Society, 128(49), 15632-15636 (2006)
Highly heat resistant and thermo-oxidatively stable borosilane alkynyl hybrid polymers
Hua Z,et al.
Royal Society of Chemistry Advances, 5, 12161-12167 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service