Skip to Content
Merck
All Photos(1)

Key Documents

295493

Sigma-Aldrich

Propyne

≥99%

Synonym(s):

Allylene, Methylacetylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C≡CH
CAS Number:
Molecular Weight:
40.06
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

204.6 mmHg ( −49.5 °C)

Assay

≥99%

bp

−23.2 °C (lit.)

mp

−102.7 °C (lit.)

SMILES string

CC#C

InChI

1S/C3H4/c1-3-2/h1H,2H3

InChI key

MWWATHDPGQKSAR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

The use of brass control valve Z146021 or brass regulator Z513539 is recommended.

hose barb

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Gas 1A - Press. Gas Compr. Gas - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

2A - Gases

WGK

nwg

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A I Strom et al.
The journal of physical chemistry. A, 124(22), 4471-4483 (2020-05-14)
Parahydrogen (pH2) quantum solids are excellent matrix isolation hosts for studying the rovibrational dynamics and nuclear spin conversion (NSC) kinetics of molecules containing indistinguishable nuclei with nonzero spin. The relatively slow NSC kinetics of propyne (CH3CCH) isolated in solid pH2
Meiyan Wang et al.
Journal of computational chemistry, 32(6), 1170-1177 (2011-03-10)
The reaction mechanism of Pd(0)-catalyzed methylacetylene bisselenation reaction is investigated by using the density functional method. The overall reaction mechanism involves the oxidative addition, insertion, and reductive elimination steps. The regioselectivity has been investigated for the methylacetylene insertion into Pd-Se
Yongping Zhang et al.
Langmuir : the ACS journal of surfaces and colloids, 29(6), 1868-1874 (2013-01-19)
To achieve silicon functionalization for the development of hybrid devices, multifunctional molecules may be employed to attach to the silicon surfaces. It is important to get a fundamental understanding about the molecule/silicon interface chemistry and the binding configuration. The surface
Michael J Ardolino et al.
Journal of the American Chemical Society, 134(21), 8770-8773 (2012-05-19)
The palladium-catalyzed cross-coupling of chiral propargyl acetates and allyl boronates delivers chiral 1,5-enynes with excellent levels of chirality transfer and can be applied across a broad range of substrates.
Changkun Li et al.
The Journal of organic chemistry, 72(19), 7431-7434 (2007-08-25)
The BF3.OEt2-catalyzed Friedel-Crafts propargylation of aromatic compounds with O-propargyl trichloroacetimidates is highly efficient and affords 1,3-diarylpropyne derivatives in good yields.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service