All Photos(2)
About This Item
Linear Formula:
H2NCH2CH2C6H4SO2NH2
CAS Number:
Molecular Weight:
200.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
99%
form
powder
mp
150-152 °C (lit.)
functional group
amine
SMILES string
NCCc1ccc(cc1)S(N)(=O)=O
InChI
1S/C8H12N2O2S/c9-6-5-7-1-3-8(4-2-7)13(10,11)12/h1-4H,5-6,9H2,(H2,10,11,12)
InChI key
FXNSVEQMUYPYJS-UHFFFAOYSA-N
Gene Information
human ... CA1(759) , CA2(760) , CA5A(763) , CA5B(11238) , CA9(768)
mouse ... Car13(71934) , Car5a(12352)
General description
The human hepatocellular carcinoma (HCC) cells were treated with its inhibitor 4-(2-aminoethyl)benzenesulfonamide.
Application
4-(2-Aminoethyl)benzenesulfonamide was used to develop a model system to study the implementation of a microfluid chip required for Fourier transform measurements of biochemical interactions.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
P Megyeri et al.
Immunology, 86(4), 629-635 (1995-12-01)
Monocytes freshly isolated from human blood produced large amounts of superoxide when triggered by phorbol ester. After monocytes were cultured for 18-24 hr in endotoxin-free, non-adherent conditions, they produced low amounts of superoxide. Addition of lipopolysaccharide (LPS), interferon-gamma (IFN-gamma), tumour
Su-jong Yu et al.
Acta pharmacologica Sinica, 32(7), 912-920 (2011-06-15)
The hypoxic condition within large or infiltrative hypovascular tumors produces intracellular acidification, which could activate many signaling pathways and augment cancer cell growth and invasion. Carbonic anhydrase-IX (CA-IX) is an enzyme lowering pH. This study is to examine whether hypoxia
Gayan S Jayawickrama et al.
PloS one, 13(4), e0196404-e0196404 (2018-04-25)
The mammalian kynurenine aminotransferase (KAT) enzymes are a family of related isoforms that are pyridoxal 5'-phosphate-dependent, responsible for the irreversible transamination of kynurenine to kynurenic acid. Kynurenic acid is implicated in human diseases such as schizophrenia where it is found
C H Mastrangelo et al.
Conference proceedings : ... Annual International Conference of the IEEE Engineering in Medicine and Biology Society. IEEE Engineering in Medicine and Biology Society. Annual Conference, 2010, 5318-5321 (2010-11-26)
New developments in microfluidic chip technology enable the construction of chemical spectrum analyzers that can probe the binding interactions between chemical entities. In this paper we report the implementation of a microfluidic chip suitable for Fourier transform measurements of biochemical
Layne D Williams et al.
Analytical chemistry, 82(14), 6025-6031 (2010-06-24)
Surface plasmon resonance (SPR) is a popular technique for label-free detection of biomolecular interactions at a surface. SPR yields quantitative kinetic association and dissociation constants of surface interactions such as the binding of two molecular species, one present in the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service