Skip to Content
Merck
All Photos(1)

Key Documents

218308

Sigma-Aldrich

α-Pinene oxide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

liquid

refractive index

n20/D 1.469 (lit.)

bp

102-103 °C/50 mmHg (lit.)

density

0.964 g/mL at 25 °C (lit.)

SMILES string

CC1(C)[C@H]2CC3OC3(C)[C@@H]1C2

InChI

1S/C10H16O/c1-9(2)6-4-7(9)10(3)8(5-6)11-10/h6-8H,4-5H2,1-3H3/t6-,7-,8?,10?/m1/s1

InChI key

NQFUSWIGRKFAHK-BGPATTHWSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hendrik Schewe et al.
Applied microbiology and biotechnology, 78(1), 55-65 (2007-12-07)
Escherichia coli BL21, expressing a quintuple mutant of P450(BM-3), oxyfunctionalizes alpha-pinene in an NADPH-dependent reaction to alpha-pinene oxide, verbenol, and myrtenol. We optimized the whole-cell biocatalyst by integrating a recombinant intracellular NADPH regeneration system through co-expression of a glucose facilitator
Mass spectrometric studies of DNA adducts from a reaction with terpenoids.
Wolfgang Schrader et al.
Angewandte Chemie (International ed. in English), 43(48), 6657-6660 (2004-12-14)
Series: 'Current issues in mutagenesis and carcinogenesis', No. 25.
P A Lefevre
Mutation research, 260(1), 5-7 (1991-05-01)
E T Griffiths et al.
Journal of bacteriology, 169(11), 4972-4979 (1987-11-01)
Over 20 gram-positive bacteria were isolated by elective culture with (+/-)-alpha-pinene as the sole carbon source. One of these strains, Nocardia sp. strain P18.3, was selected for detailed study. alpha-Pinene-grown cells oxidized, without lag, alpha-pinene, alpha-pinene oxide (epoxide), and the
Denis Linares et al.
Bioresource technology, 99(11), 4590-4596 (2007-09-15)
The feasibility of trans-2-methyl-5-isopropylhexa-2,5-dienoic acid (novalic acid) accumulation using the alpha-pinene degradation pathway of Pseudomonas rhodesiae CIP 107491 was studied. This appeared possible by using concentrated living bacterial cells produced under oxygen limitation with alpha-pinene as sole carbon source. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service