Skip to Content
Merck
All Photos(2)

Key Documents

217735

Sigma-Aldrich

5-Amino-4,6-dichloropyrimidine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3Cl2N3
CAS Number:
Molecular Weight:
163.99
Beilstein:
126885
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

mp

145-148 °C (lit.)

solubility

95% ethanol: soluble 50 mg/mL, clear to slightly hazy, colorless to faintly yellow

storage temp.

2-8°C

SMILES string

Nc1c(Cl)ncnc1Cl

InChI

1S/C4H3Cl2N3/c5-3-2(7)4(6)9-1-8-3/h1H,7H2

InChI key

NIGDWBHWHVHOAD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-Amino-4,6-dichloropyrimidine was used in the synthesis of:
  • oxepane ring containing monocyclic, conformationally restricted bicyclic and spirocyclic nucleosides
  • conformationally locked bicyclo[2.2.1]heptane/oxa-bicyclo[3.2.1]octane nucleosides
  • N(7)-substituted purines
  • chiral derivatives of (+)-erythro-9-(2-hydroxy-3-nonyl)adenine
  • 9-alkyl-6-substituted-purine analogs, potent anticonvulsant agents
  • pyrimido-oxazepines in a three-step process with microwave heating at 150°C

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Subhankar Tripathi et al.
The Journal of organic chemistry, 72(19), 7427-7430 (2007-08-25)
Carbohydrate-derived substrates having (i) C-5 nitrone and C-3-O-allyl, (ii) C-4 vinyl and a C-3-O-tethered nitrone, and (iii) C-5 nitrone and C-4-allyloxymethyl generated tetracyclic isoxazolidinooxepane/-pyran ring systems upon intramolecular nitrone cycloaddition reactions. Deprotection of the 1,2-acetonides of these derivatives followed by
Jinglin Liu et al.
Journal of combinatorial chemistry, 8(3), 410-416 (2006-05-09)
A regiospecific strategy for the preparation of N(7)-substituted purines in an efficient manner was devised. This approach to 6,7,8-trisubstituted purines relies on the cyclization reactions of suitably substituted pyrimidines (1) with either a carboxylic acid or an aldehyde. The method
J L Kelley et al.
Journal of medicinal chemistry, 31(3), 606-612 (1988-03-01)
Several 9-alkyl-6-substituted-purines were synthesized and tested for anticonvulsant activity against maximal electroshock-induced seizures (MES) in rats. Most compounds were prepared in three steps from 5-amino-4,6-dichloropyrimidine or in two steps via alkylation of 6-chloropurine. Potent anticonvulsant activity against MES resided in
Tetrahedron Letters, 48, 1489-1489 (2007)
Sk Sahabuddin et al.
The Journal of organic chemistry, 71(16), 5980-5992 (2006-07-29)
The carbohydrate-derived substrate 3-C-allyl-1,2:5,6-di-O-isopropylidene-alpha-D-allofuranose was judiciously manipulated for preparing suitable synthons, which could be converted to a variety of isoxazolidino-spirocycles and -tricycles through the application of ring-closing metathesis (RCM) and intramolecular nitrone cycloaddition (INC) reactions. Cleavage of the isoxazolidine rings

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service