Skip to Content
Merck
All Photos(2)

Key Documents

202150

Sigma-Aldrich

Cesium nitrate

99.99% trace metals basis

Synonym(s):

Nitric acid cesium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CsNO3
CAS Number:
Molecular Weight:
194.91
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99% trace metals basis

form

powder

impurities

≤150.0 ppm Trace Metal Analysis

mp

414 °C (lit.)

density

3.685 g/mL at 25 °C (lit.)

SMILES string

[Cs+].[O-]N(=O)=O

InChI

1S/Cs.NO3/c;2-1(3)4/q+1;-1

InChI key

NLSCHDZTHVNDCP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in the synthesis of cesium nitratocuprate, Cs2[Cu(NO3)4], which crystallizes in a square-planar coordination around the Cu center.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Ox. Sol. 1 - Repr. 2

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Morozov, I.V. et al.
Zeitschrift fur Anorganische und Allgemeine Chemie, 624, 1543-1543 (1998)
Alex M James et al.
ACS applied materials & interfaces, 11(25), 22464-22473 (2019-05-30)
Sulfonated hyper-cross-linked polymers based on 4,4'-bis(chloromethyl)-1,1'-biphenyl (BCMBP) were synthesized via metal-free (SHCP-1) and conventional Lewis acid-catalyzed (SHCP-2) Friedel-Crafts alkylation routes. The sulfonated polymers possessed BET surface areas in excess of 500 m2·g-1. SHCP-1 was investigated for its ability to extract
Morozov, I.V. et al.
Zeitschrift fur Anorganische und Allgemeine Chemie, 624, 1543-1543 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service