Skip to Content
Merck
All Photos(2)

Key Documents

199583

Sigma-Aldrich

Lissamine Green B

Dye content 60 %

Synonym(s):

Acid Green 50, Wool Green S

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H25N2NaO7S2
CAS Number:
Molecular Weight:
576.62
Colour Index Number:
44090
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

Quality Level

composition

Dye content, 60%

solubility

H2O: 0.1%

λmax

633 nm

ε (extinction coefficient)

≥10000 at 300-306 nm in water at 0.005 g/L
≥50000 at 236-242 nm in water at 0.005 g/L
≥60000 at 630-636 nm in water at 0.005 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].CN(C)c1ccc(cc1)C(=C2/C=CC(\C=C2)=[N+](/C)C)\c3c(O)c(cc4cc(ccc34)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C27H26N2O7S2.Na/c1-28(2)20-9-5-17(6-10-20)25(18-7-11-21(12-8-18)29(3)4)26-23-14-13-22(37(31,32)33)15-19(23)16-24(27(26)30)38(34,35)36;/h5-16H,1-4H3,(H2,31,32,33,34,35,36);/q;+1/p-1

InChI key

WDPIZEKLJKBSOZ-UHFFFAOYSA-M

Application

Lissamine Green B has been used in blood-brain barrier (BBB) studies and is used to study the its effect on herpes simplex virus 1 (HSV-1) corneal infection in animals.

Biochem/physiol Actions

Lissamine green also referred to as acid green 50, wool green S, food green S or green S, is used for vital staining of cornea and conjunctiva. Staining characteristics are almost similar to rose bengal dye. Lissamine green is also used to stain membrane-damaged epithelial cells. In vitro, Lissamine green B at low concentration blocks herpes simplex virus 1 (HSV-1) plaque formation.

Legal Information

Lissamine is a trademark of PerkinElmer Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photocatalytic degradation of Lissamine Green B dye by using nanostructured sol?gel TiO2 films
Curkovic L
J. Alloy Compounds, 604, 309-316 (2014)
Hannah Dueck et al.
Genome biology, 16, 122-122 (2015-06-10)
Differentiation of metazoan cells requires execution of different gene expression programs but recent single-cell transcriptome profiling has revealed considerable variation within cells of seeming identical phenotype. This brings into question the relationship between transcriptome states and cell phenotypes. Additionally, single-cell
Vascular Permeability and Microcirculation of Gliomas and Mammary Carcinomas
Transplanted in Rat and Mouse Cranial Windows1
Yuan F
Cancer Research, 54, 4564- 4568 (1994)
Donald R Korb et al.
Cornea, 29(4), 377-383 (2010-02-20)
The purpose of this study was to investigate the prevalence of lid wiper epitheliopathy (LWE) in patients diagnosed with dry eye disease (DED). Patients were recruited for two groups. Inclusion criteria for the DED group (n = 50) was: a
S Rockwell et al.
Experimental gerontology, 28(3), 281-293 (1993-05-01)
Radiobiological data and measurements with O2 microelectrodes show that EMT6 tumors implanted into aged mice have a higher proportion of radioresistant, hypoxic cells than do tumors implanted into young adult animals; radiation is less effective in killing cells in tumors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service