Skip to Content
Merck
All Photos(3)

Documents

178098

Sigma-Aldrich

Glutarimide

98%

Synonym(s):

2,6-Piperidinedione, NSC 58190

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7NO2
CAS Number:
Molecular Weight:
113.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

155-157 °C (lit.)

SMILES string

O=C1CCCC(=O)N1

InChI

1S/C5H7NO2/c7-4-2-1-3-5(8)6-4/h1-3H2,(H,6,7,8)

InChI key

KNCYXPMJDCCGSJ-UHFFFAOYSA-N

General description

A glutarimide antibiotic, 9-methylstreptimidone, shows antiviral, antitumor and antifungal activities.

Application

Reactant for:
Thionations
Biocatalytic asymmetric synthesis of chiral amines from ketones applied to sitagliptin manufacture
Synthesis of β-adrenoceptor ligands
Enantioselective synthesis of securinega alkaloids
Intramolecular amidocyclopropanation reactions
Synthesis of alpha-fluoro-alpha amino amides

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Cai-Yun Geng et al.
Journal of computational chemistry, 29(5), 686-693 (2007-09-13)
The reaction mechanism of the Rh-catalyzed [4 + 2] annulation of 4-alkynals with isocyanates is unraveled using density functional calculations. The reaction mechanisms of the model system and the real substituted system have been investigated and the results are compared.
Yohsuke Watanabe et al.
Organic letters, 12(2), 268-271 (2009-12-22)
The boron aldol reaction of beta-substituted glutaric imides bearing an oxazolidinone-based auxiliary proceeds with excellent diastereoselectivity; switching the tertiary amine employed between i-Pr(2)EtN or Et(3)N affords enantiomeric lactone product.
Matthew Hoffmann et al.
Cancer chemotherapy and pharmacology, 71(2), 489-501 (2012-12-04)
To investigate the pharmacokinetics and disposition of [(14)C]pomalidomide following a single oral dose to healthy male subjects. Eight subjects were administered a single 2 mg oral suspension of [(14)C]pomalidomide. Blood (plasma), urine and feces were collected. Mass balance of radioactivity
Hung-Wei Chen et al.
Organic letters, 8(14), 3033-3035 (2006-06-30)
[reaction: see text] The reaction of alpha-sulfonyl acetamide 1 with various cyclic unsaturated esters 2 to fused bicyclic glutarimides is reported. Syntheses of (+/-)-alloyohimbane (4) and louisianin D (5) have been accomplished.
Jianhua Ju et al.
Journal of the American Chemical Society, 127(34), 11930-11931 (2005-08-25)
Iso-Migrastatin (10) has been shown to be the main natural product of Streptomyces platensis, which undergoes a facile, H2O-mediated rearrangement into dorrigocin A (2), 13-epi-dorrigocin A (11), dorrigocin B (3), and migrastatin (1). Eight new congeners (12-19) of 10 were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service