Skip to Content
Merck
All Photos(4)

Key Documents

155721

Sigma-Aldrich

Methacrylic acid

contains 250 ppm MEHQ as inhibitor, 99%

Synonym(s):

2-Methacrylic acid, 2-Methylpropenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)COOH
CAS Number:
Molecular Weight:
86.09
Beilstein:
1719937
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

>3 (vs air)

Quality Level

vapor pressure

1 mmHg ( 20 °C)

Assay

99%

form

liquid

autoignition temp.

752 °F

contains

250 ppm MEHQ as inhibitor

refractive index

n20/D 1.431 (lit.)

pH

2.0-2.2 (20 °C, 100 g/L)

bp

163 °C (lit.)

mp

12-16 °C (lit.)

density

1.015 g/mL at 25 °C (lit.)

SMILES string

C=C(C)C(O)=O

InChI

1S/C4H6O2/c1-3(2)4(5)6/h1H2,2H3,(H,5,6)

InChI key

CERQOIWHTDAKMF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methacrylic acid (MAA) is an organic compound, colorless, viscous liquid. It is a monofunctional methacrylayte monomer. It is soluble in warm water and miscible in most organic solvents. MAA is used as a starting material for the production of its esters, especially methyl methacrylate and poly methyl methacrylate (PMMA).

Application

MAA is used to synthesize poly(methacrylic acid)-g-poly(ε-caprolactone) copolymers, polymeric blends with PMMA and hydroxyethyl methacrylate (HEMA)-MAA hydrogels.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reversible Addition Fragmentation Chain Transfer
(RAFT) Polymerization
Moad G, et al.
Material Matters, 5(1) (2010)
Synthesis of graft copolymers of poly(methacrylic acid)-g-poly(?-caprolactone) by coupling ROP and RAFT polymerizations
Kiehl J, et al.
Polymer, 53(3), 694-700 (2012)
Thomas J Dursch et al.
Biomaterials, 35(2), 620-629 (2013-10-24)
Two-photon confocal microscopy and back extraction with UV/Vis-absorption spectrophotometry quantify equilibrium partition coefficients, k, for six prototypical drugs in five soft-contact-lens-material hydrogels over a range of water contents from 40 to 92%. Partition coefficients were obtained for acetazolamide, caffeine, hydrocortisone
Josué A Torres-Ávalos et al.
Polymers, 13(4) (2021-03-07)
Design of a smart drug delivery system is a topic of current interest. Under this perspective, polymer nanocomposites (PNs) of butyl acrylate (BA), methacrylic acid (MAA), and functionalized carbon nanotubes (CNTsf) were synthesized by in situ emulsion polymerization (IEP). Carbon
Riteshkumar J Patel et al.
Journal of biomedical materials research. Part A, 100(8), 2054-2062 (2012-05-15)
Exudate analysis indicated that both hydroxamated methacrylic acid-based (HX-MAA) and methacrylic acid-based (MAA) beads without hydroxamation, implanted in an air pouch with gelatin discs, elicited similar biological responses, with both resulting in high cell counts and chemokine (MCP3) levels, relative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service