Skip to Content
Merck
All Photos(1)

Documents

141070

Sigma-Aldrich

Isopropyl isocyanate

≥98%

Synonym(s):

1-Methylethyl isocyanate, 2-Isocyanatopropane, Isopropyl isocyanate, Methyl ethyl isocyanate, Propan-2-yl isocyanate, n-Isopropyl isocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHNCO
CAS Number:
Molecular Weight:
85.10
Beilstein:
969356
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98%

form

liquid

refractive index

n20/D 1.382 (lit.)

bp

74-75 °C (lit.)

density

0.866 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)N=C=O

InChI

1S/C4H7NO/c1-4(2)5-3-6/h4H,1-2H3

InChI key

GSLTVFIVJMCNBH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Isopropyl isocyanate was used as reagent during the synthesis of O-aryl N-isopropylcarbamates. It was used as derivatization reagent during the stereoisomeric analysis of secondary alcohols.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

26.6 °F - closed cup

Flash Point(C)

-3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service