Skip to Content
Merck
All Photos(2)

Key Documents

110051

Sigma-Aldrich

5-Ethyl-2-methylpyridine

≥96%

Synonym(s):

5-Ethyl-2-picoline, NSC 1984

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11N
CAS Number:
Molecular Weight:
121.18
Beilstein:
109269
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.2 (vs air)

Assay

≥96%

refractive index

n20/D 1.497 (lit.)

bp

178 °C (lit.)

density

0.919 g/mL at 25 °C (lit.)

SMILES string

CCc1ccc(C)nc1

InChI

1S/C8H11N/c1-3-8-5-4-7(2)9-6-8/h4-6H,3H2,1-2H3

InChI key

NTSLROIKFLNUIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-Ethyl-2-methylpyridine is one of the components that contribute to the nutty, roasted aroma in the Parmigiano-Reggiano cheese.

Application

5-Ethyl-2-methylpyridine, associated with Maillard reaction chemistry and sugar thermal degradation processes, has been used to study the nanogram adsorption for SPME headspace and liquid sampling.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M Qian et al.
Journal of dairy science, 85(6), 1362-1369 (2002-07-31)
The volatile compounds in Parmigiano-Reggiano cheese were isolated by solvent extraction/high vacuum distillation, followed by separation into acidic, basic, water-soluble, and neutral fractions. The neutral fraction was further fractionated by a normal-phase liquid chromatography. Aroma compounds were identified using gas
A Study of the Behavior of Maillard Reaction Products Analyzed by Solid-Phase Microextraction?Gas Chromatography?Mass Selective Detection.
Coleman WM
Journal of Chromatographic Science, 34(5), 213-218 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service