Mesoporous Silica Applications
John P. Hanrahan, Tom F. O’Mahony, Joseph M. Tobin, John J. Hogan
Glantreo Ltd ERI Building, Lee Road, Cork City, Ireland
Background
Since their discovery in the late 1970s, mesoporous silica have attracted attention because of their unique properties, such as ordered pore structures, high specific surface areas, and their possible synthesis in a wide range of morphologies such as spheres rods, discs, and powders. Unlike traditional porous silica, mesoporous silica exhibit exceptionally ordered pores. This arises from the nanotemplating approach applied during synthesis of these materials. Over the years, a plethora of mesoporous silica (SBA 15, SBA 16, MCM 41, MCM 48, etc.) with a wide range of pore geometries (hexagonal, cubic, etc.) and particle morphologies, such as discs, spheres and rods have been synthesised. Figure 1 shows some of the morphologies of mesoporous silica (MS) and porous silica spheres (PSS). The porous silica spheres are spherical particles with quasi ordered porosity. Recent developments in the manufacturing of mesoporous silica, allowing large-scale production (up to kg scale), has facilitated their move from laboratory based research to more advanced application driven research.
Mesoporous silica with uniform and tailorable pore dimensions with high surface areas are being used many applications, including wastewater remediation, indoor air cleaning, catalysis, bio-catalysis, drug delivery, CO2 capture, bioanalytical sample preparation, pervaporation membrane improvement. Mesoporous silica are also used as templates for controlling the aspect ratio of quantum-confined nanoparticles and nanowires. Table 1 summarizes some of the application areas and thematic areas of PSS and MS.
![Scanning electron microscopy (SEM) and transmission electron microscopy (TEM) images showing varying morphologies and pore structures of porous silica spheres (PSS) (A&B) and mesoporous silica (MS) (C&D). Scanning electron microscopy and transmission electron microscopy (TEM) images of porous silica spheres](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/environmental-testing-and-industrial-hygiene/waste-water-and-process-water-testing/sem-tem-porous-silica-spheres/sem-tem-porous-silica-spheres.jpg)
Figure 1.Scanning electron microscopy (SEM) and transmission electron microscopy (TEM) images showing varying morphologies and pore structures of porous silica spheres (PSS) (A&B) and mesoporous silica (MS) (C&D).
Application | Thematic Area | Material Type / Product |
---|---|---|
Quantum confined nanowire host | Electronics/Energy | SBA-15 (Product No. 806803, 806854, 806862) |
Heavy metal ion removal from waste water | Environmental | SBA-15 (Product No. 806803, 806854, 806862), SBA-16 (Product No. 806927), MCM-48 (Product No. 805467) |
Phosphate removal from waste water | Environmental | Metal (e.g. titanium) doped SBA-15 (Product No. 806919) |
Removal of indoor volatile organic compounds (VOCs) | Environmental/Health | PSS |
Methanolysis of styrene oxide | Catalysis | Metal (e.g. titanium) doped SBA-15 (Product No. 806919) |
Improving the bioavailability of poorly water soluble drug molecules | Drug Delivery/Pharmaceutical formulation | SBA-15 (Product No. 806803, 806854, 806862) |
Enzyme encapsulation for bio catalysis | Catalysis | SBA-15(Product No. 806803, 806854, 806862) /PSS |
Phospholipid extraction from biological matrices | Bio-Analysis/Sample Preparation | Metal doped SBA-15 (Product No. 806919) |
Incorporation of spherical silica particles in pervaporation membranes for the separation of water from ethanol | Chemical Separation | PSS |
Direct air capture of CO2 by physisorbent materials | Energy | SBA-15 (Product No. 806803, 806854, 806862) |
Mesoporous Silica as Quantum Confined Nanowire Hosts
Because of their unique optical, electrical, and mechanical properties, the nanoscale structures of semiconductor wires are expected to play a vital role in emerging technologies. MS that contain unidirectional arrays of pores, typically 2-15 nm in diameter, running throughout the material have been successfully exploited as templates for semiconductor materials formed from the gas phase. Recently, a novel supercritical fluid solution-phase based approach to produce silicon nanowires within the pores of mesoporous silica was also reported.1
![X-ray diffraction (XRD) and TEM image of Si wires grown inside the pores of MS. The XRD and TEM images of the MS before (TOP) and after (BOTTOM) its pores were filled with silicon nanowires. The high packing density of the silicon “mesowires” in the silica matrix makes them ideally suited for the formation of quantum structures. X-ray diffraction (XRD) and TEM image of Si wires grown inside the pores of MS](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/environmental-testing-and-industrial-hygiene/waste-water-and-process-water-testing/xrd-tem-silica-wires/xrd-tem-silica-wires.jpg)
Figure 2.X-ray diffraction (XRD) and TEM image of Si wires grown inside the pores of MS. The XRD and TEM images of the MS before (TOP) and after (BOTTOM) its pores were filled with silicon nanowires. The high packing density of the silicon “mesowires” in the silica matrix makes them ideally suited for the formation of quantum structures.
Heavy Metal Ion Removal from Waste Water Using Modified Mesoporous Silica
Metal ions are a significant environmental pollutant in wastewater, and long-term exposure to solvated metal ions and their effects on human health and natural ecosystems is a major concern. The metals of primary interest include chromium, nickel, manganese, iron and various heavy metals. Several methods, such as precipitation, coagulation/flocculation, ion exchange, reverse osmosis, complexation/sequestration, electrochemical operation and biological treatment are used for removal of metal ions from waste. However, these methods have limitations from high operating and energy costs. Sorbents, such as activated charcoal, zeolites and clays have also been for wastewater treatment . However, disadvantages of these materials include relatively low and variable loading capacities and small metal ion-binding constants. Owing to their large surface areas (typically 200–1,000 cm2 g−1), large pore volumes and ease of recycling/regeneration, the ordered MS are becoming established sorptives for metals ions. In fact, the functionalization of mesoporous silica with various chelating agents (or metal ion-specific ligands) allows selective removal of metal ions from aqueous or organic systems at larger uptakes. Table 2 shows the sorption capacities for the mono-functionalised (amine or thiol) and bi-functionalized (amine and thiol) SBA-15.2
Initial Concentration (µmol/g) | Si-NH | Si-SH | SH-Si-NH | |
---|---|---|---|---|
Zn | 152.9 ± 0.2 | 0.6 ± 0.4 | 30.8 ± 5.1 | 1.3 ± 1 |
Pb | 48.3 ± 0.1 | 0 ± 1.1 | 7.7 ± 1.8 | 1.3 ± 0.8 |
Cd | 88.9 ± 0.1 | 0.8 ± 0.3 | 13.1 ± 1 | 0.1 ± 0.3 |
Ni | 170.4 ± 0.2 | 16 ± 3.7 | 30.1 ± 4.1 | 2.4 ± 3.1 |
Fe | 179.1 ± 0.2 | 1.6 ± 2.2 | 17.3 ± 3.9 | 0.2 ± 0.1 |
Cr | 192.3 ± 0.3 | 18.9 ± 0.9 | 26.5 ± 1.4 | 3.2 ± 2.8 |
Mg | 411.4 ± 0.5 | 338.1 ± 88.1 | 75.2 ± 12.1 | 0 ± 0.2 |
V | 196.3 ± 0.3 | 7 ± 4.2 | 6.7 ± 1.6 | 0 ± 0.3 |
Mn | 182 ± 0.3 | 3.5 ± 4.7 | 27.6 ± 1 | 0 ± 0.1 |
Ca | 249.5 ± 0.4 | 93 ± 24.2 | 25 ± 2.1 | 3.8 ± 1.2 |
Cu | 157.4 ± 0.2 | 0 ± 1.8 | 0 ± 0.3 | 0 ± 0.2 |
Phosphate Removal from Waste Water Using Metal Doped Mesoporous Silica
Phosphorus (as phosphate ions) is an important element that is widely used in agriculture as a fertilizer and in industry as a detergent. However, the use of phosphorus causes eutrophication when it is released into aquatic environments. Several Environmental Protection Agencies worldwide have identified eutrophication resulting from excess phosphorus input as the greatest threat to fresh water supply. Eutrophication leads to fish deaths and the degradation of habitat with loss of plant and animal species. Ordered MS have been proven to be a good adsorbent for metal ions and amino acids. Recently there has been significant focus on using MS functionalized with selective ligands for metal ion adsorption. Also, MS doped with transition metals have been shown to be promising for phosphate adsorption. Figure 3 outlines several doped mesoporous silica and their phosphate removal efficiencies from model aqueous solutions. In all cases, the doped SBA-15 material exhibit high adsorption efficiency.3
![Phosphate removal from waste water using titanium, iron, zirconium and aluminium doped SBA-15. Several doped mesoporous silica and their phosphate removal efficiencies from model aqueous solutions](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/environmental-testing-and-industrial-hygiene/waste-water-and-process-water-testing/phospate-removal-waste-water.gif)
Figure 3.Phosphate removal from waste water using titanium, iron, zirconium and aluminium doped SBA-15.
Removal of Volatile Organic Carbons (VOCs) from Indoor Air Using Porous Silica Spheres (PSS)
Indoor air pollution has an adverse effect on human health. The health effects of indoor air pollution are expected to increase. In 2001, the national human activity pattern survey found that in the United States people typically spend ~90% of their time indoors. Aldehydes are known to have adverse health effects (eye and lung irritation), and formaldehyde and acrolein are suspected carcinogens. Changes in building design and improved energy efficiency, along with increase in insulation and reduction in air exchange, have led to increasingly airtight buildings. Modern synthetic building materials, such as sealants, plastics and solvent-based coatings have added to the problem and volatile organic compounds (VOCs), non-volatile organic compounds (NVOCs) and semi-volatile organic compounds (SVOCs) are indoor pollutants.
Porous silica spheres (PSS) can be used to efficiently trap various indoor air pollutants both in simulated and indoor environments. The adsorbent was tested at relatively high concentrations (500 ppb) and flow rates (10 L min-1). It was shown that PSS outperforms the commercially available Amberlite XAD-4 resin at trapping non-polar VOCs and it is significantly more efficient at trapping polar VOCs present in ambient air. PSS adsorbent trapped 100% of the gas phase carbonyl compounds present in a simulation chamber experiment in the first 10 min of sampling, and the XAD-4 resin had various levels of efficiency ranging from 100 to 8% over the sampling period for the same group of carbonyl compounds.
Figure 4 shows a trapping efficiency plot for a selection of small carbonyls: acetone, butanal, pentanal and hexanal. The trapping efficiency of the PSS for each of these compounds is close to 100% after the first 10 min. The XAD-4 resin showed varying trapping efficiency values ranging from 100 to 8% after the first 10 min. However, the trapping efficiency for both sorbents decreased gradually with time due to the progressive saturation of the sorption surface with trapped species.4
![Data showing the trapping efficiency of XAD-4 (dashed line) and PSS (solid line) for (a) acetone, (b) butanal, (c) pentanal and (d) hexanal. Trapping efficiency of XAD-4 and PSS for acetone, butanal,pentanal and (d) hexanal](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/environmental-testing-and-industrial-hygiene/waste-water-and-process-water-testing/trapping-efficiency-xad4-pss.gif)
Figure 4.Data showing the trapping efficiency of XAD-4 (dashed line) and PSS (solid line) for (a) acetone, (b) butanal, (c) pentanal and (d) hexanal.
Metal Doped Mesoporous Silica for the Methanolysis of Styrene Oxide
Highly ordered zirconium and titanium doped hexagonal MS with Si/Zr and Si/Ti ratios of 40:1 and 80:1 have been used as solid acid catalysts for the methanolysis of styrene oxide in a single-mode microwave reactor. As catalyst Si/Zr material demonstrated excellent substrate conversion, high product selectivity and remained highly active for several reaction cycles (Figure 5). These experimental results show that the zirconium doped mesoporous silica is an efficient catalyst for the liquid-phase methanolysis of styrene oxide. The isolation of the products required a simple filtration/evaporation step. A typical reaction carried out with a 40:1 Si:Zr catalyst in a single-cavity microwave reactor operating at 105 W for 10 min afforded 100% conversion of styrene oxide. Recycling studies have shown that the catalyst remains highly efficient for at least five cycles with 95% conversion of styrene oxide after the catalyst was subjected to microwave irradiation while suspended in methanol.5
![(TEM) image of 40.1 Si/Zr sample showing hexagonal ordering and pore retention (TEM) image of 40.1 Si/Zr sample showing hexagonal ordering and pore retention](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/environmental-testing-and-industrial-hygiene/waste-water-and-process-water-testing/tem-si-zr-hexagonal-ordering.gif)
![Transmission electron microscopy (TEM) image of 40.1 Si/Zr sample showing hexagonal ordering (A) and pore retention (B). Bottom image shows the percentage styrene oxide conversion vs Recyclability and total turnover number (TON) for 40:1 catalyst (bottom image) percentage styrene oxide conversion vs Recyclability and total turnover number (TON) for 40:1 catalyst](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/environmental-testing-and-industrial-hygiene/waste-water-and-process-water-testing/tem-si-zr-pore-retention.gif)
Figure 5.Transmission electron microscopy (TEM) image of 40.1 Si/Zr sample showing hexagonal ordering (A) and pore retention (B). Bottom image shows the percentage styrene oxide conversion vs Recyclability and total turnover number (TON) for 40:1 catalyst (bottom image)
Drug/SBA-15 Formulations for Improving the Bioavailability of Poorly Water Soluble Drug Molecules
It is well established that increasing the effective surface area of a poorly water-soluble drug in contact with the dissolution medium can enhance drug dissolution. This can be achieved by loading drugs onto ordered MS, which are characterized by high surface areas, large mesopore volumes, narrow mesopore size distributions (5–8 nm), and ordered unidirectional mesopore networks. These properties allow the homogeneous and reproducible drug-loading and release with the use of MS.
![SEM/EDX images of physical mix of fenofibrate loaded SBA-15 SEM/EDX images of physical mix of fenofibrate loaded SBA-15](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/environmental-testing-and-industrial-hygiene/waste-water-and-process-water-testing/sem-edx-images-fenofibrate-sba-15/sem-edx-images-fenofibrate-sba-15.jpg)
![SEM/EDX images of physical mix of fenofibrate loaded SBA-15 (top image) and melt sample (bottom). The SEM images are on the left, and the EDX one are on the right. Image A in the bottom part shows the release profiles of unprocessed fenofibrate, physical mix and melt samples whereas the release profile of impregnated, liquid and SC-CO2 processed samples are shown in image B. SEM/EDX images of physical mix of fenofibrate loaded SBA-15](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/environmental-testing-and-industrial-hygiene/waste-water-and-process-water-testing/sem-edx-images-melt-sample/sem-edx-images-melt-sample.jpg)
Figure 6.SEM/EDX images of physical mix of fenofibrate loaded SBA-15 (top image) and melt sample (bottom). The SEM images are on the left, and the EDX one are on the right. Image A in the bottom part shows the release profiles of unprocessed fenofibrate, physical mix and melt samples whereas the release profile of impregnated, liquid and SC-CO2 processed samples are shown in image B.
The release of drug from the silica carrier is a key performance indicator to consider when employing OMMs (ordered mesoporous materials) for drug dissolution enhancement. The in vitro release of drug from drug–silica samples and the dissolution of the starting fenofibrate are shown in Figure 6. Using MS as a carrier material improved the drug dissolution rate for all processed samples.6
Enzyme Encapsulation into Mesoporous Silica for Biocatalysis
Immobilization of enzymes can confer a number of advantages such as enhanced stability, ease of recovery and re-use, and the ability of using the enzyme in non-aqueous solvents where the enzyme is insoluble. However, the main disadvantage of immobilization is that it usually lowers the activity of the enzyme and the process of immobilization can add significant costs to the process. In addition, immobilization methods tend to be non-specific and typically the process of immobilization of a specific enzyme on a support is optimized and developed on a case-by-case basis. Support materials for the immobilization of an enzyme should be mechanically and chemically stable, have high surface area, be easily made at low cost and display low non-specific protein adsorption properties. Also, the immobilization should occur in a manner which does not compromise the enzyme conformation or activity, while maintaining the diffusion of the substrate and product to and from the active site. MS has been widely used as supports for enzymes, and significant efforts have been made to use them as supports for biocatalysis.
![Activity profiles for cytochrome c immobilised on SBA-15 (Square), PSS (Triangle), and for aqueous cytochrome c (open circles). Activity profiles for cytochrome c immobilised on SBA-15, PSS, and for aqueous cytochrome c](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/environmental-testing-and-industrial-hygiene/waste-water-and-process-water-testing/activity-profiles-cytochrome-c-sba-15/activity-profiles-cytochrome-c-sba-15.jpg)
Figure 7.Activity profiles for cytochrome c immobilised on SBA-15 (Square), PSS (Triangle), and for aqueous cytochrome c (open circles).
The adsorption and activity of cytochrome c and lipase on two silica supports (SBA-15 and PSS) of the same average pore diameter but with different pore volumes and surface areas has been examined. On SBA-15 loadings of cytochrome c and Candida antarctica lipase B (CALB) of 15.6 and 2.04 mol g−1 were obtained in comparison to loadings of 0.94 and 6.7 mol g−1 on PPS respectively. These differences in loading can be ascribed to differences in the properties (pore volume, surface area and morphology) of the supports. The catalytic activity of cytochrome c was similar on both supports, while the activity of CALB was higher on SBA-15 in comparison to PPS (7.8 vs 4 s−1). These differences in activity for CALB likely arise from the pore morphology and the physical properties of the supports, which is the hypothesis that was supported by the higher recyclability obtained with SBA 15. The data indicates that the physical properties of mesoporous silica supports can significantly alter the activity and stability of enzymes.7
Doped Mesoporous Silica for Phospholipid Extraction from Biological Matrices
Food matrices are difficult to work with because they contain unwanted or interfering compounds (i.e. phospholipids), and samples often require a clean-up and extraction before determination using liquid chromatography (LC) and/or mass spectroscopy. Recently doped MS (SBA-15 doped with titanium) was used in dispersive solid phase extraction (dSPE) QuEChERS (quick, easy, cheap, effective, rugged and safe) based approach to sample preparation of liver tissues. Titanium doped SBA-15 has been used for preparation of biological samples before high performance liquid chromatography (HPLC) analysis and it has been shown to have preferential selectivity for phospholipids without removing the analyte of interest from the sample. The doping of the titanium moiety into the silica framework offers significant advantages in terms of robustness and chemical stability over other metal silica hybrid materials where the metal moieties are attached to pure silica.
The recoveries from samples processed with the titanium doped mesoporous silica (SiTi) are higher than those processed with traditional C18 sorbent material. The data presented in Table 3 for C18 and SiTi(4%)-C18 sorbent materials suggest that SiTi(4%)-C18 is more effective than traditional C18 sorbent used in QuEChERS based method for anthelmintics sample preparation in d-SPE format.
Analyte | C18 Mean (n=5) | ±SD | CV% | SiTi4%C18 Mean (n=5) | ±SD | CV% |
---|---|---|---|---|---|---|
Albendazole | 80 | 12 | 15 | 125 | 10 | 8 |
Albendazole-sulphoxide | 71 | 5 | 8 | 99 | 6 | 6 |
Albendazole-sulphone | 81 | 5 | 6 | 121 | 5 | 4 |
Albendazole-amino-sulphone | 61 | 1 | 1 | 78 | 6 | 8 |
Cambendazole | 81 | 4 | 5 | 112 | 8 | 7 |
Fenbendazole | 83 | 10 | 12 | 123 | 9 | 8 |
Oxfendazole | 72 | 4 | 6 | 109 | 7 | 7 |
Fenbendazole-sulphone | 77 | 5 | 7 | 125 | 12 | 10 |
Flubendazole | 78 | 5 | 7 | 121 | 8 | 7 |
Amino-flubendazole | 54 | 2 | 4 | 76 | 6 | 8 |
Hydroxy-flubendazole | 62 | 3 | 5 | 90 | 6 | 7 |
Mebendazole | 76 | 6 | 8 | 124 | 9 | 7 |
Amino-mebendazole | 50 | 1 | 3 | 71 | 5 | 6 |
Hydroxy-mebendazole | 77 | 6 | 7 | 116 | 7 | 6 |
Oxibendazole | 85 | 9 | 11 | 119 | 9 | 8 |
Triclabendazole | 88 | 15 | 17 | 138 | 10 | 7 |
Triclabendazole-sulphoxide | 77 | 6 | 7 | 107 | 1 | 1 |
Triclabendazole-sulphone | 93 | 6 | 7 | 143 | 2 | 1 |
Thiabendazole | 82 | 3 | 3 | 97 | 3 | 3 |
5-hydroxy-thiabendazole | 67 | 3 | 5 | 83 | 7 | 8 |
Levamisole | 78 | 2 | 2 | 96 | 5 | 6 |
Clorsulon | 79 | 5 | 6 | 111 | 0 | 0 |
Closantel | 92 | 10 | 11 | 135 | 7 | 5 |
Morantel | 75 | 1 | 2 | 101 | 7 | 7 |
Niclosamide | 90 | 14 | 15 | 124 | 8 | 6 |
Nitroxynil | 59 | 3 | 5 | 79 | 1 | 1 |
Oxyclozanide | 85 | 9 | 10 | 107 | 3 | 3 |
Rafoxanide | 87 | 18 | 21 | 139 | 9 | 7 |
Monepantel | 99 | 10 | 10 | 128 | 6 | 4 |
Monepantel Sulphone | 95 | 8 | 9 | 127 | 6 | 4 |
Abamectin B1a | 100 | 23 | 23 | 137 | 2 | 1 |
Doramectin | 93 | 18 | 19 | 133 | 6 | 4 |
Emamectin B1a | 78 | 11 | 14 | 132 | 8 | 6 |
Eprinomectin B1a | 79 | 18 | 23 | 175 | 10 | 6 |
Ivermectin B1a | 89 | 11 | 13 | 117 | 6 | 5 |
Moxidectin | 89 | 13 | 15 | 157 | 8 | 5 |
Overall | 79 | 8 | 9 | 116 | 6 | 6 |
Incorporation of Porous Silica Spheres (PSS) into Pervaporation Membranes for the Separation of Water from Ethanol
Pervaporation is a membrane separation technology primarily used to dehydrate and recover solvents. It is also used to separate organic–organic mixtures. The pervaporation technology has significant advantage over other separation techniques as it can be used to effectively ‘break’ azeotropes of mixtures without any physical difficulties. The technique also does not suffer from the negative environmental impacts of techniques like azeotropic distillation. One method of improving membrane flux without significantly compromising selectivity is by inclusion of porous particles (e.g. zeolites and silica particles) into the polymer matrix. In porous ceramic–polymer membrane hybrids of this type the engineering of the particles (i.e. size, shape, monodispersity, pore size and surface chemistry) is of significant importance.
![Absorption of 50:50 wt% solution of ethanol and water into selective membrane over time Absorption of 50:50 wt% solution of ethanol and water into selective membrane over time](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/environmental-testing-and-industrial-hygiene/waste-water-and-process-water-testing/membrane-absorption-vs-time-chart/membrane-absorption-vs-time-chart.jpg)
![Membrane absorption vs. time: the upper image shows the absorption of 50:50 wt% solution of ethanol and water into selective membrane over time. The maxima in the graph represents point at which membrane becomes dissolute i.e. loses membrane form and becomes unusable. The picture A in the bottom part shows the surface image of 15 wt% silica loaded PVA membrane whereas the image B shows the surface image of 10 wt% silica loaded PVA membrane. surface image of 15 wt vs 10 wt silica loaded PVA membrane](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/environmental-testing-and-industrial-hygiene/waste-water-and-process-water-testing/membrane-absorption-vs-time-surface-image/membrane-absorption-vs-time-surface-image.jpg)
Figure 8.Membrane absorption vs. time: the upper image shows the absorption of 50:50 wt% solution of ethanol and water into selective membrane over time. The maxima in the graph represents point at which membrane becomes dissolute i.e. loses membrane form and becomes unusable. The picture A in the bottom part shows the surface image of 15 wt% silica loaded PVA membrane whereas the image B shows the surface image of 10 wt% silica loaded PVA membrane.
The incorporation of engineered PSS into polymer pervaporation membranes can be highly beneficial. For example, it has been demonstrated that incorporation of spherical and discreet sized monodispersed silica particles of 1.8–2 μm diameter with a pore size of 1.8 nm into a poly (vinyl alcohol) [PVA] polymer produces composite pervaporation membrane that exhibits significant increases in both flux and selectivity (Figure 8). Unlike zeolitic systems, PSS can be controllably engineered to give a wide range of pore sizes and chemistries and may provide new generations membranes for various pervaporation applications.8
Direct Air Capture of CO2 using Modified Mesoporous Silica
Generation of carbon dioxide (CO2) poses a significant threat to global climate and it represents a topical challenge that requires attention. In this context, the anthropogenic emissions of CO2 are a concern as it is generated as an undesirable component of commodities like natural gas, biogas, etc. However, it is argued that sequestration of CO2 either from gas mixtures or directly from air could mitigate the risks associated with carbon emissions and various porous materials such as mesoporous silica, zeolites, metal organic frameworks have been examined for this purpose. Recent work has shown that amine functionalized SBA-15 out performs the benchmark materials like metal organic frameworks (MOFs) and zeolite in CO2 sequestration.9
Network error: Failed to fetch