HPLC Analysis of Tamoxifen E/Z isomers and Related Compounds on Ascentis® Express RP-Amide
CONDITIONS
column
Ascentis Express RP-Amide, 10 cm x 3 mm I.D., 2.7 μm particles (53918-U)
mobile phase
[A] 0.1% formic acid in water; [B] 0.1% formic acid in acetonitrile
gradient
held at 25% B for 2 min; 25 to 40% B in 15 min; 40 to 25% B in 0.1 min; held at 25% B for 2.9 min
flow rate
0.6 mL/min
pressure
2292 psi (158 bar)
column temp.
35 °C
detector
UV, 260 nm
injection
2 μL
sample
10 mg/L in 10:90, water: methanol
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany