Merck
  • Levonorgestrel, medroxyprogesterone and progesterone cause a concentration-dependent reduction in endometrial cancer (Ishikawa) cell density, and high concentrations of progesterone and mifepristone act in synergy.
Levonorgestrel, medroxyprogesterone and progesterone cause a concentration-dependent reduction in endometrial cancer (Ishikawa) cell density, and high concentrations of progesterone and mifepristone act in synergy.
Anticancer research (2009-05-06)
Bjørn T Moe, Anne B Vereide, Anne Orbo, Ragnhild Jaeger, Georg Sager
ABSTRACT
Endometrial hyperplasia is a precursor lesion of endometrial carcinoma. Clinical studies of endometrial hyperplasia have shown that levonorgestrel (LNG) is more therapeutically effective than medroxyprogesterone acetate (MPA). The present pharmacological in vitro study was performed to compare progestin effects on human endometrial cancer (Ishikawa) cells. Supraphysiological concentrations of progesterone (PG) and high concentrations of LNG and MPA were employed to determine the order of potency in reducing cell density. The order of potency was LNG>MPA>PG with respective 50% inhibitory concentrations (IC(50)) of 3.9+/-0.4, 30.4+/-3.4 and 45.3+/-2.7 microM. Mifepristone (MF) is a potent antiprogestin, but was unable to antagonize the PG-induced cell density reduction. For MF concentrations from 0.2 to 70 microM alone, a PG-mimetic effect was observed with an IC(50) value of 19.0+/-1.7 muM. When PG and MF were combined, a marked reinforcement of the effect was seen. These observations indicate that extranuclear initiated signaling pathways are involved in the reduction of endometrial cancer cells exposed to high concentrations of PG and MF.
Product Description
Supelco
ORBO 402 Tenax® TA specially treated (35/60), 100/50 mg, W,W,W separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 609 Amberlite XAD®-2 (20/50) 400/200 mg, W,W,W separators, O.D. × L 8 mm × 110 mm, pkg of 50 ea
Supelco
ORBO 53 Activated Silica Gel (20/40) specially cleaned, 400/200 mg with Glass Fiber Filters, GFF,F,F separators, O.D. × L 7 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 502 Activated Silica Gel (20/45) 100/50 mg, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 49P (OVS) Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 270/140 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS) Tube, pkg of 10 ea
Supelco
ORBO 554 H2SO4 on Silica Gel (20/40) 150/75 mg, W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 80 Coated Filter, with cassettes unassembled
Supelco
ORBO 23 2-HMP on Amberlite XAD®-2 (20/40), 120/60 mg, W,W,W separators, O.D. × L 6 mm × 85 mm, pkg of 25 ea
Supelco
ORBO 80 Isocyanate Filter, with cassettes: no, filter O.D. 37 mm, pkg of 25 ea
Supelco
ORBO 52 Small Activated Silica Gel (20/40) 150/75 mg, pk 50, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 24 2-HMP on Amberlite XAD®-2 (20/40), 150/75 mg, W,W,W separators, O.D. × L 6 mm × 105 mm, pkg of 25 ea
Supelco
ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
Supelco
ORBO 92 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group Vinyl acetate
Supelco
ORBO 65P (OVS) Amberlite XAD®-4 specially cleaned, 160/80 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS), pkg of 10 ea
Supelco
ORBO 43 Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 100/50 mg, W,W,W (specially treated) separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea