Merck
  • Activating de novo mutations in NFE2L2 encoding NRF2 cause a multisystem disorder.
Activating de novo mutations in NFE2L2 encoding NRF2 cause a multisystem disorder.
Nature communications (2017-10-12)
Peter Huppke, Susann Weissbach, Joseph A Church, Rhonda Schnur, Martina Krusen, Steffi Dreha-Kulaczewski, W Nikolaus Kühn-Velten, Annika Wolf, Brenda Huppke, Francisca Millan, Amber Begtrup, Fatima Almusafri, Holger Thiele, Janine Altmüller, Peter Nürnberg, Michael Müller, Jutta Gärtner
ABSTRACT
Transcription factor NRF2, encoded by NFE2L2, is the master regulator of defense against stress in mammalian cells. Somatic mutations of NFE2L2 leading to NRF2 accumulation promote cell survival and drug resistance in cancer cells. Here we show that the same mutations as inborn de novo mutations cause an early onset multisystem disorder with failure to thrive, immunodeficiency and neurological symptoms. NRF2 accumulation leads to widespread misregulation of gene expression and an imbalance in cytosolic redox balance. The unique combination of white matter lesions, hypohomocysteinaemia and increased G-6-P-dehydrogenase activity will facilitate early diagnosis and therapeutic intervention of this novel disorder.The NRF2 transcription factor regulates the response to stress in mammalian cells. Here, the authors show that activating mutations in NRF2, commonly found in cancer cells, are found in four patients with a multisystem disorder characterized by immunodeficiency and neurological symptoms.
Sigma-Aldrich
Anti-NFE2L2 antibody produced in rabbit
Sigma-Aldrich
Luteolin, ≥98% (TLC), powder
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid