Merck
  • Secreted gelsolin desensitizes and induces apoptosis of infiltrated lymphocytes in prostate cancer.
Secreted gelsolin desensitizes and induces apoptosis of infiltrated lymphocytes in prostate cancer.
Oncotarget (2017-11-05)
Chun-Chi Chen, Shiow-Her Chiou, Cheng-Lin Yang, Kuan-Chih Chow, Tze-Yi Lin, Hui-Wen Chang, Weir-Chiang You, Hisu-Wen Huang, Chien-Min Chen, Nien-Cheng Chen, Fen-Pi Chou, Ming-Chih Chou
ABSTRACT
Loss of immunosurveillance is a major cause of cancer progression. Here, we demonstrate that gelsolin, a constituent of ejaculate, induces apoptosis of activated lymphocytes in prostate cancer. Gelsolin was highly expressed in prostate cancer cells, and was associated with tumor progression, recurrence, metastasis, and poor prognosis. In vitro, secreted gelsolin inactivated CD4+ T cells by binding to CD37, and induced apoptosis of activated CD8+ T lymphocytes by binding to Fas ligand during cell contact dependent on major histocompatibility complex I. Moreover, secreted gelsolin bound to sortilin, which in turn bound to Wiskott-Aldrich syndrome protein family member 3, thereby enhancing the endocytosis and intracellular transport of essential lipids needed to facilitate tumor growth and expansion. Under normal conditions, gelsolin is a seemingly harmless protein that prevents immune responses in female recipients. In disease states, however, this protein can inhibit immunosurveillance and promote cancer progression.
Sigma-Aldrich
1,1′-Carbonyl-di-(1,2,4-triazole), technical, ≥90% (T)
Sigma-Aldrich
Neurotensin, ≥90% (HPLC)
Sigma-Aldrich
trans-Dehydroandrosterone, ≥99%
Sigma-Aldrich
Human EGFR / ErbB1 ELISA Kit, for serum, plasma, cell culture supernatants and urine
Sigma-Aldrich
Anti-SORT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution