Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Home
Search Results
[Ag
[Ag
Nanoscale (2017-06-06)
Esma Khatun, Atanu Ghosh, Debasmita Ghosh, Papri Chakraborty, Abhijit Nag, Biswajit Mondal, Sudhakar Chennu, Thalappil Pradeep
PMID
28581554
ABSTRACT
We report the synthesis of a new silver cluster, [Ag
Product Description
269093
Sigma-Aldrich
1,3-Benzenedithiol, 99%
Pricing
F15315
Sigma-Aldrich
4-Fluorothiophenol, 98%
Pricing
125237
Sigma-Aldrich
4-Chlorothiophenol, 97%
Pricing
275441
Sigma-Aldrich
2,4-Dimethylbenzenethiol, 95%
Pricing