Merck
  • Salivary IgA and IgG antibodies to bullous pemphigoid 180 noncollagenous domain 16a as diagnostic biomarkers in mucous membrane pemphigoid.
Salivary IgA and IgG antibodies to bullous pemphigoid 180 noncollagenous domain 16a as diagnostic biomarkers in mucous membrane pemphigoid.
The British journal of dermatology (2015-12-18)
S Ali, C Kelly, S J Challacombe, A N A Donaldson, J K G Dart, M Gleeson, J F Setterfield
ABSTRACT
Mucous membrane pemphigoid (MMP) is an uncommon mucocutaneous immunobullous disorder. Use of saliva for diagnosis by enzyme-linked immunosorbent assay (ELISA) using the noncollagenous (NC) domain 16a of bullous pemphigoid antigen II (BP180) is not well described. To establish whether whole or parotid saliva is a suitable alternative to serum for diagnosis of MMP. Precoated BP180-NC16a ELISA plates were used to test serum, and whole and parotid saliva for IgG, IgA and secretory IgA antibodies. Patients with MMP (n = 64) provided matched serum and whole saliva. In addition 18 of the MMP patients also provided matched parotid saliva. Healthy controls (n = 50) provided matched serum and whole saliva and 6 of these additionally provided matched parotid saliva. An additional 16 disease controls provided matched serum, and whole and parotid saliva. In whole saliva, IgG antibodies were detected in 11/64 (17%), IgA in 23/64 (36%) and a combined positivity in 29/64 (45%). In parotid saliva, IgA antibodies were found in 8/18 (44%). Serum IgG antibodies were detected in 27/64 (42%), serum IgA antibodies in 18/64 (28%) and a combined positivity in 33/64 (52%). Combined use of serum and saliva increased detection of specific antibodies by 30%. Control samples were all negative (positive predictive value of 100% for all tests). The negative predictive values were 62% for IgA saliva, 65% for IgG serum, 59% for IgA serum and 56% for IgG saliva. IgG and IgA antibodies may provide a suitable diagnostic marker in MMP. Assay of salivary IgA antibodies to NC16a offers a similar diagnostic predictive value to serum.
Sigma-Aldrich
Monoclonal Anti-Secretory Component (IgA) antibody produced in mouse, clone GA-1, ascites fluid
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Human IgA (α-chain specific)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution