Merck
  • Increased internalization of complement inhibitor CD59 may contribute to endothelial inflammation in obstructive sleep apnea.
Increased internalization of complement inhibitor CD59 may contribute to endothelial inflammation in obstructive sleep apnea.
Science translational medicine (2016-01-08)
Memet Emin, Gang Wang, Francesco Castagna, Josanna Rodriguez-Lopez, Romina Wahab, Jing Wang, Tessa Adams, Ying Wei, Sanja Jelic
ABSTRACT
Obstructive sleep apnea (OSA), characterized by intermittent hypoxia (IH) during transient cessation of breathing, triples the risk for cardiovascular diseases. We used a phage display peptide library as an unbiased approach to investigate whether IH, which is specific to OSA, activates endothelial cells (ECs) in a distinctive manner. The target of a differentially bound peptide on ECs collected from OSA patients was identified as CD59, a major complement inhibitor that protects ECs from the membrane attack complex (MAC). A decreased proportion of CD59 is located on the EC surface in OSA patients compared with controls, suggesting reduced protection against complement attack. In vitro, IH promoted endothelial inflammation predominantly via augmented internalization of CD59 and consequent MAC deposition. Increased internalization of endothelial CD59 in IH appeared to be cholesterol-dependent and was reversed by statins in a CD59-dependent manner. These studies suggest that reduced complement inhibition may mediate endothelial inflammation and increase vascular risk in OSA patients.
Sigma-Aldrich
Anti-MCAM Antibody, clone P1H12, biotin conjugated, clone P1H12, Chemicon®, from mouse