Merck
  • Aspergillus mulundensis sp. nov., a new species for the fungus producing the antifungal echinocandin lipopeptides, mulundocandins.
Aspergillus mulundensis sp. nov., a new species for the fungus producing the antifungal echinocandin lipopeptides, mulundocandins.
The Journal of antibiotics (2015-10-16)
Gerald F Bills, Qun Yue, Li Chen, Yan Li, Zhiqiang An, Jens C Frisvad
ABSTRACT
The invalidly published name Aspergillus sydowii var. mulundensis was proposed for a strain of Aspergillus that produced new echinocandin metabolites designated as the mulundocadins. Reinvestigation of this strain (Y-30462=DSMZ 5745) using phylogenetic, morphological, and metabolic data indicated that it is a distinct and novel species of Aspergillus sect. Nidulantes. The taxonomic novelty, Aspergillus mulundensis, is introduced for this historically important echinocandin-producing strain. The closely related A. nidulans FGSC A4 has one of the most extensively characterized secondary metabolomes of any filamentous fungus. Comparison of the full-genome sequences of DSMZ 5745 and FGSC A4 indicated that the two strains share 33 secondary metabolite biosynthetic gene clusters. These shared gene clusters represent ~45% of the total secondary metabolome of each strain, thus indicating a high level intraspecific divergence in terms of secondary metabolism.
Sigma-Aldrich
N-Z-Amine® EKC, Casein enzymatic hydrolysate, from bovine milk
Pricing and availability is not currently available.