Merck
  • Lymphatic endothelial cells are a replicative niche for Mycobacterium tuberculosis.
Lymphatic endothelial cells are a replicative niche for Mycobacterium tuberculosis.
The Journal of clinical investigation (2016-02-24)
Thomas R Lerner, Cristiane de Souza Carvalho-Wodarz, Urska Repnik, Matthew R G Russell, Sophie Borel, Collin R Diedrich, Manfred Rohde, Helen Wainwright, Lucy M Collinson, Robert J Wilkinson, Gareth Griffiths, Maximiliano G Gutierrez
ABSTRACT
In extrapulmonary tuberculosis, the most common site of infection is within the lymphatic system, and there is growing recognition that lymphatic endothelial cells (LECs) are involved in immune function. Here, we identified LECs, which line the lymphatic vessels, as a niche for Mycobacterium tuberculosis in the lymph nodes of patients with tuberculosis. In cultured primary human LECs (hLECs), we determined that M. tuberculosis replicates both in the cytosol and within autophagosomes, but the bacteria failed to replicate when the virulence locus RD1 was deleted. Activation by IFN-γ induced a cell-autonomous response in hLECs via autophagy and NO production that restricted M. tuberculosis growth. Thus, depending on the activation status of LECs, autophagy can both promote and restrict replication. Together, these findings reveal a previously unrecognized role for hLECs and autophagy in tuberculosis pathogenesis and suggest that hLECs are a potential niche for M. tuberculosis that allows establishment of persistent infection in lymph nodes.
Sigma-Aldrich
Mannan from Saccharomyces cerevisiae, prepared by alkaline extraction
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
Saponin, used as non-ionic surfactant
Sigma-Aldrich
ECO TWEEN® 80, viscous liquid
Millipore
Middlebrook 7H9 Broth Base, suitable for microbiology, NutriSelect® Plus,
Recommended for use in isolation and cultivation of Mycobacterium species
Sigma-Aldrich
Fibronectin bovine plasma, solution, sterile-filtered, BioReagent, suitable for cell culture
Millipore
Skim Milk Powder, suitable for microbiology
Millipore
Middlebrook 7H11 Agar Base, suitable for microbiology, NutriSelect® Plus
Sigma-Aldrich
TWEEN® 80, viscous liquid