Merck
  • Effect of molecular characteristics on the formation of nitrosamines during chlor(am)ination of phenylurea herbicides.
Effect of molecular characteristics on the formation of nitrosamines during chlor(am)ination of phenylurea herbicides.
Environmental science. Processes & impacts (2015-11-06)
Wei-Hsiang Chen, Ya-Ciao Yang, Ya-Hong Wang, Chi-Min Li, Kun-Yi Lin, Jie-Chung Lou
ABSTRACT
The objective of this study was to investigate the formation of different nitrosamines during chlorination or chloramination (chlor(am)ination) of five phenylurea herbicides (fluometuron, diuron, linuron, metobromuron, and propanil), with the effects of disinfection approaches, additional inorganic nitrogen, and reaction pH being studied. By analyzing six nitrosamines, N-nitrosodimethylamine (NDMA) and N-nitrosopyrrolidine (NPYR) formation was observed. The dimethylamine functional group was the key to determining whether a particular phenylurea herbicide was an important nitrosamine precursor, as the NDMA conversion ratio was much higher. Chlorination with ammonium or dichloramination enhanced the NDMA formation. NPYR formation from the herbicides that did not form NDMA was detected and was more vigorous during dichloramination or in the presence of either ammonium or nitrite. The NPYR formation was possibly related to the aniline molecular fragment from the phenylurea herbicides. Both NDMA and NPYR formation were higher at pH 8. Overall, the maximum nitrosamine conversions decreased in the order: fluometuron > diuron > propanil > metobromuron > linuron (up to 0.99%, 0.46%, 0.005%, 0.004%, and 0.003% molar conversion rates, respectively) during chlorination or chloramination and dichloramine > free chlorine > monochloramine (up to 0.99%, 0.41%, and 0.005% molar conversion rates, respectively) for given herbicide, chlorine, and nitrogen doses. Applying the results of this study, phenylurea herbicide concentrations ranging from several to tens of μg L(-1) will yield a NDMA concentration in drinking water above the level for a theoretical 10(-6) lifetime cancer risk. NPYR formation will increase the risk of these phenylurea herbicide concentrations to downstream water users. The true adverse environmental impacts of these phenylurea herbicides are important to emphasize given their high loadings as non-point source pollutants and their typical environmental scenarios (e.g., at neutral pH or with the co-occurrence of inorganic nitrogen), likely resulting in more efficient nitrosamine formation.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Potassium nitrate, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Ammonium chloride, for molecular biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Diuron, ≥98%
Sigma-Aldrich
Potassium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Ammonium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Potassium nitrate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium chloride, 99.998% trace metals basis
Sigma-Aldrich
Potassium nitrate, 99.99% trace metals basis
Sigma-Aldrich
Ammonium chloride, 99.99% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Sodium hypochlorite solution, purum, ~10% (RT)
Sigma-Aldrich
Potassium nitrate, puriss., meets analytical specification of Ph Eur, BP, USP, FCC, E252, 99.0-100.5%
Sigma-Aldrich
Ammonium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium nitrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium hypochlorite solution, 6-14% active chlorine basis
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Potassium nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Ammonium chloride, ReagentPlus®, ≥99.5%