Merck
  • Direct metabolomics for plant cells by live single-cell mass spectrometry.
Direct metabolomics for plant cells by live single-cell mass spectrometry.
Nature protocols (2015-08-28)
Takashi Fujii, Shuichi Matsuda, Mónica Lorenzo Tejedor, Tsuyoshi Esaki, Iwao Sakane, Hajime Mizuno, Naohiro Tsuyama, Tsutomu Masujima
ABSTRACT
Live single-cell mass spectrometry (live MS) provides a mass spectrum that shows thousands of metabolite peaks from a single live plant cell within minutes. By using an optical microscope, a cell is chosen for analysis and a metal-coated nanospray microcapillary tip is used to remove the cell's contents. After adding a microliter of ionization solvent to the opposite end of the tip, the trapped contents are directly fed into the mass spectrometer by applying a high voltage between the tip and the inlet port of the spectrometer to induce nanospray ionization. Proteins are not detected because of insufficient sensitivity. Metabolite peaks are identified by exact mass or tandem mass spectrometry (MS/MS) analysis, and isomers can be separated by combining live MS with ion-mobility separation. By using this approach, spectra can be acquired in 10 min. In combination with metabolic maps and/or molecular databases, the data can be annotated into metabolic pathways; the data analysis takes 30 min to 4 h, depending on the MS/MS data availability from databases. This method enables the analysis of a number of metabolites from a single cell with rapid sampling at sub-attomolar-level sensitivity.
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
DL-Tyrosine, 99%
Sigma-Aldrich
Perchloric acid, 70%, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Acetonitrile