Merck
  • Color Tuning of the Aggregation-Induced Emission of Maleimide Dyes by Molecular Design and Morphology Control.
Color Tuning of the Aggregation-Induced Emission of Maleimide Dyes by Molecular Design and Morphology Control.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-07-17)
Hiroaki Imoto, Kohei Kizaki, Seiji Watase, Kimihiro Matsukawa, Kensuke Naka
ABSTRACT
Aggregation-induced emission (AIE)-active maleimide dyes, namely, 2-p-toluidino-N-p-tolylmaleimide, 3-phenyl-2-toluidino-N-p-tolylmaleimide, 2-p-thiocresyl-3-p-toluidino-N-p-tolylmaleimide, and 2,3-dithiocresyl-N-arylmaleimides, were synthesized by facile synthetic procedures. The dyes show intense emission in the solid state, and emission colors were controlled from green (λmax =527 nm) to orange (λmax =609 nm) by varying the substituents at the 2- and 3-positions of the maleimide and the packing structures in the solid state. 2,3-Disubstituted maleimide dyes effectively underwent redshifts of their emission wavelength. Furthermore, some of the dyes exhibited mechanochromism and polymorphism, and their emission properties were dramatically dependent on the morphology of the solid samples. The mechanisms of the emission behaviors were investigated by X-ray diffraction. The substituent of the nitrogen atom of the maleimide ring affected the intermolecular interactions and short contacts, which were observed by single crystal X-ray crystallography, to result in completely different emission properties.
Sigma-Aldrich
Bis(triphenylphosphine)palladium(II) dichloride
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Triethylamine, purum, ≥99% (GC)
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Bis(triphenylphosphine)palladium(II) dichloride, 98%
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
p-Anisidine, 99%
Sigma-Aldrich
Maleic anhydride, 99%
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Bis(triphenylphosphine)palladium(II) dichloride
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Phenylboronic acid, 95%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
4-Methylbenzenethiol, 98%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
p-Toluidine, 99.6%
Sigma-Aldrich
p-Toluidine, 99%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Toluene, anhydrous, 99.8%